Tubing Connectors
Bel-Art™ SP Scienceware™ Laboratory Tubing Fittings Assortment Kit Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Fittings are for the most common lab connections and compatible with tubing measuring 3.2 to 12.7mm (⅛ to ½ in.)
Fisherbrand™ Y-Shaped Connectors with Straight Ends
Autoclavable and highly resistant to many chemicals
Thermo Scientific™ Nalgene™ Valved Barbed Quick Disconnects Available on GSA/VA Contract for Federal Government customers only.
Maintain the integrity of your transfer liquids using Thermo Scientific™ Nalgene™ Valved Barbed Quick Disconnects.
Thermo Scientific™ Dry-Bags™ Tubing Connector
Use this stainless steel connector to attach Thermo Scientific™ peristaltic pump tubing to the filling tubing that is part of each Thermo Scientific™ Dry-Bag™ .
Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors Available on GSA/VA Contract for Federal Government customers only.
Maximize liquid flow with non-tapered hose barbed Y-Type Connectors, molded of polypropylene resin, free of animal derived components.
Fisherbrand™ Tubing Four-Way Polypropylene Connectors Products for Science Education
Work in any tubing or vacuum system.
Thermo Scientific™ Nalgene™ HDPE Quick Disconnects Available on GSA/VA Contract for Federal Government customers only.
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ Quick Disconnects.
Fisherbrand™ Tubing Y Polypropylene Connectors Products for Science Education
Work in any tubing or vacuum system.
Fisherbrand™ Tubing L Polypropylene Connectors Products for Science Education
Work in any tubing or vacuum system.
Thermo Scientific™ Nalgene™ Tubing T-Type Connectors Available on GSA/VA Contract for Federal Government customers only.
Maximize liquid flow with non-tapered hose barbed T-Type Connectors, molded of polypropylene resin, free of animal derived components.
Fisherbrand™ Polypropylene Tubing 4-Way Connectors
Very durable, autoclavable, and highly resistant to many chemicals
Fisherbrand™ Tubing T Connectors Products for Science Education
Work in any tubing or vacuum system.
Thermo Scientific™ Nalgene™ Sanitary 1 in. Hose Barb Connector Available on GSA/VA Contract for Federal Government customers only.
Nalgene Sanitary Tri-clamp Hose barb fitting supports connections between 1 ″ sanitary fitting to flexible tubing with inner diameter of 0.5″.
Bel-Art™ SP Scienceware™ All-Metal Quick-Disconnects
Inline hose connector with automatic shutoff on disconnect to prevent leakage
Bel-Art™ SP Scienceware™ T Connectors Available on GSA/VA Contract for Federal Government customers only.
Connectors have multiple barbs per arm providing additional contact between the fitting and the tubing for extra gripping power
Bel-Art™ SP Scienceware™ 5-in-1 Connectors Available on GSA/VA Contract for Federal Government customers only.
Fits 5 sizes of tubing and will connect the same size tubing on both ends, or will act as a reducer for any combination within the range
Wheaton™ Hose Connector Available on GSA/VA Contract for Federal Government customers only.
Grease-free fitting for condensers and connection to a vacuum
Bel-Art™ SP Scienceware™ Complete Fitting Assortment Available on GSA/VA Contract for Federal Government customers only.
Reduces time spent searching for the right tubing connector or stopper
Fisherbrand™ Polyethylene Quick-Disconnects
For easy joining of plastic or rubber tubing
Bel-Art™ SP Scienceware™ Reducing Quick-Disconnects Available on GSA/VA Contract for Federal Government customers only.
Each consists of two barbed polyethylene connectors which assemble tightly together with a male-female center taper
  spinner