Weighing Dishes
Fisherbrand™ Polystyrene Weighing Dishes Products for Science Education
Ideal for weighing, dispensing or storing applications.
Fisherbrand™ Polystyrene Antistatic Weighing Dishes
Disposable dishes are ideal for weighing, dispensing, or storing applications
Fisherbrand™ Aluminum Weighing Dishes
Aluminum dishes for weighing and dispensing
Fisherbrand™ Disposable Aluminum Weighing/Drying Pans
Designed for weighing, drying and more. Compatible with many popular balances and moisture analyzers. Choice of 10.9cm and 12.2cm diameter dishes.
Fisherbrand™ Disposable Aluminum Dishes with Fluted Sides
Useful as evaporating and general utility dishes
Mettler Toledo™ Moisture Analyzer Sample Pans Available on GSA/VA Contract for Federal Government customers only.
For halogen and infrared moisture analyzers
Qorpak™ Aluminum Weigh Dishes with Tab Available on GSA/VA Contract for Federal Government customers only.
Perfect for routine weighing, moisture testing and embedding mold
Fisherbrand™ Hexagonal Polystyrene Weighing Dishes
Design provides greater balance, protection and safety
Mettler Toledo™ Aluminum Weigh Boats for Micro and Ultramicro Series Available on GSA/VA Contract for Federal Government customers only.
For discontinued Micro (MT5) and Ultramicro (UMT2) Series balances
Qorpak™ Disposable Aluminum Moisture Balance Pans Available on GSA/VA Contract for Federal Government customers only.
Made from oil free aluminum to provide even temperatures across each pan
Whatman™ Grade 2122 Weighing Papers Available on GSA/VA Contract for Federal Government customers only.
Constructed of Pergamyne paper. Whatman™ Grade 2122 Weighing Papers provide excellent quantitative transfer from paper. Shiny surface confirms complete transfer.
Whatman™ Kjeldahl Analysis Weighing Boats, Grade 609 Available on GSA/VA Contract for Federal Government customers only.
Designed to collect, transfer, and drop both sample and weighing boat into the acid solution during a Kjeldahl analysis without influencing analytical results. Whatman™ Kjeldahl Weighing Boats are excellent for both weighing and quantitative sample transfer.
Sartorius™ Aluminum Weighing Boats Available on GSA/VA Contract for Federal Government customers only.
Variety of sizes available
Sartorius™ Polystyrene Weighing Boats Available on GSA/VA Contract for Federal Government customers only.
Made of anti-static polystyrene
Dyn-A-Med Weighing Canoes
Resist dilute acids, aqueous solutions, alcohols and bases
Disposable Polystyrene Weighing Dishes Products for Science Education
Easy-grip contour sides for nonslip handling.
Disposable Aluminum Pan Liners Products for Science Education
A convenient and inexpensive solution for easier cleanups.
Disposable Aluminum Dishes Products for Science Education
Withstand temperatures up to 677°C (1250.6°F).
Sartorius™ Infrared Moisture Analyzer Disposable Foil Sample Dishes
For use with all sartorius Moisture Analyzers
  spinner