Distillation Columns Traps Tubes
PYREX™ Vacuum-Type Distillation Traps Available on GSA/VA Contract for Federal Government customers only.
Separable, with standard taper joints
PYREX™ Dean-Stark Trap Available on GSA/VA Contract for Federal Government customers only.
For water from petroleum or bitumens with volatile solvents
Kimble™ Kontes™ Distilling Trap Adapter, Foam-Type
Place in boiling flask to prevent foaming-type reactions from entering either the distilling column or the condenser
PYREX™ Barrett Trap Available on GSA/VA Contract for Federal Government customers only.
For Barrett test to determine water content in petroleum or bituminous products
Kimble™ Kontes™ Brand Distilling Columns
Used with Kontes Evaporative Concentrators
Kimble™ Kontes™ Micro Condenser Distillation Columns
Supplied jacket allows column to be used as a condenser
Kimble™ Kontes™ Claisen Distillation Adapter
Provides dual entry into a variety of flasks
PYREX™ Distilling Adapter Tube with Side Arm and standard taper Joints Available on GSA/VA Contract for Federal Government customers only.
Adapter with standard taper 24/40 outer joint at top and standard taper 24/40 inner joint at bottom
Corning™ Glass Distilling Columns Available on GSA/VA Contract for Federal Government customers only.
Widely used in the lab, especially for fractionation
Foss™ DU100 Distillation Unit and Kjeltec 2100/2200 Distillation Systems Accessories
For Fisher Scientific Model DU100 and Kjeltec™ Models 2100/2200
  spinner