Specific Gravity Bottles
Kimble™ KIMAX™ Specific Gravity Bottles, Pycnometer Available on GSA/VA Contract for Federal Government customers only.
Flask with side tube and cap plus thermometer
Kimble™ KIMAX™ Specific Gravity Bottle for Cement, Sand Available on GSA/VA Contract for Federal Government customers only.
Used to obtain specific gravity of cement, sand, other fine materials
PYREX™ Gay-Lussac Bottles Available on GSA/VA Contract for Federal Government customers only.
For determining liquid densities
PYREX™ Specific Gravity Bottle for Viscous Fluids Available on GSA/VA Contract for Federal Government customers only.
For specific gravity testing with viscous fluids, semisolid bitumens, and emulsions
Kimble™ KIMAX™ Specific Gravity Bottle for Oils, Tars Available on GSA/VA Contract for Federal Government customers only.
Determines specific gravity of various oils, tars, cements, varnishes
  spinner