Evaporating Dishes
Fisherbrand™ Evaporating Dishes Products for Science Education
Useful for a variety of evaporating processes and ashing.
Alfa Aesar™ Glassy Carbon Evaporating Dish
Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.
PYREX™ Glass Evaporating Dishes Products for Science Education
Rugged beaded rims prevent liquids from dripping.
PYREX™ Evaporating Dishes Available on GSA/VA Contract for Federal Government customers only.
Withstand temperatures to 230°C (446°F)
EISCO Porcelain Evaporating Basins Products for Science Education
Consumables for use in chemical laboratories during experiments. EISCO Evaporating Basin is used for holding samples during evaporation.
Bel-Art™ SP Scienceware™ Evaporating Dish Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polypropylene. Bel Art™ SP Scienceware™ Evaporating Dish is useful for a variety of laboratory tasks.
United Scientific Supplies Porcelainware Starter Kit Products for Science Education
Kit includes top-selling porcelain products.
BrandTech™ VITLAB™ PFA Evaporating Dishes Available on GSA/VA Contract for Federal Government customers only.
Highly resistant to heat and chemical contamination
American Educational Products Evaporating Dishes Products for Science Education
Sturdy and corrosion-resistant.
GSC International Porcelain Evaporating Dishes Products for Science Education
Designed to withstand high temperatures with proper use. GSC International Porcelain Evaporating Dishes can withstand temperatures of up to 1050°C on their glazed portions, and up to 1350°C on their unglazed bottom surfaces.
CoorsTek™ Porcelain Evaporating Dishes
Glazed except on rim for easy cleaning
CoorsTek™ Porcelain Evaporating Dishes Products for Science Education
Glazed except on rim for easy cleaning.
CoorsTek™ Porcelain Casserole Evaporating Dishes
Withstand temperatures to 1150°C (2102°F)
  spinner