Glass Volumetric Flasks
Kimble™ Kontes™ Wide Mouth Heavy Wall Class A Volumetric Flasks Available on GSA/VA Contract for Federal Government customers only.
Heavy wall design increases flask durability
Kimble™ KIMAX™ Flasks with Graduation Line Etched through Vertical Red Stripe Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Contain” within Class A tolerances
Kimble™ KIMAX™ Class B Flasks with standard taper Pennyhead Stopper Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Contain” within Class B tolerances
Kimble™ KIMAX™ Heavy-Duty RAY-SORB™ Volumetric Class A Flasks without Stoppers Available on GSA/VA Contract for Federal Government customers only.
RAY-SORB processing protects flask contents from short-length lightwaves
Kimble™ KIMAX™ Volumetric Flasks with Polyethylene Snap Caps Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Borosilicate glass flasks supplied with snap caps.
PYREX™ Flasks for Determining Phosphoric Acid in Fertilizer Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Contain,” ±0.18mL limit of error
Kimble™ BEAKERplus™ Combination Beakers and Flasks
Combination design to reduce splashing during agitation with funnels or filters
PYREX™ Class B Flasks with standard taper Stopper Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Contain” at 20°C (68°F)
PYREX™ Flasks Serialized and Certified with standard taper Barrelhead Stopper Available on GSA/VA Contract for Federal Government customers only.
Calibrated to Class A tolerances in accordance with ASTM™ E-542 and E-288
PYREX™ VISTA™ Volumetric Flasks Available on GSA/VA Contract for Federal Government customers only.
Necks are tooled for standard taper glass stoppers
PYREX™ VISTA™ Class B Flasks Available on GSA/VA Contract for Federal Government customers only.
Capacity tolerances are twice those specified for Class A volumetric ware
Lenz Laborglasinstrumente™ PYREX™ Serialized and Certified Flasks, Low Actinic Available on GSA/VA Contract for Federal Government customers only.
Featuring red low actinic stained glass to provide protection for materials sensitive to light. Lenz Laborglasinstrumente™ PYREX™ Serialized and Certified Flasks, Low Actinic are tested and calibrated in an ISO/IEC 17025 certified laboratory.
  spinner