Thistle Tube Funnels
Fisherbrand™ Classic Glass Thistle Tube Funnels Products for Science Education
Borosilicate glass with thistle top
Bel-Art™ SP Scienceware™ Thistle Tube Available on GSA/VA Contract for Federal Government customers only.
Makes additions to burets and narrow neck containers easily
  spinner