Dropping Pipets
Thermo Scientific™ Nalgene™ One-Piece Disposable LDPE Droppers Available on GSA/VA Contract for Federal Government customers only.
Manufacture 30 to 35 drops per mL with the bellows-type bulb of the Thermo Scientific™ Nalgene™ One-Piece Disposable LDPE Dropper.
Thermo Scientific™ Samco™ Graduated Transfer Pipettes Available on GSA/VA Contract for Federal Government customers only.
Accurately measure more than one volume by using these graduated polyethylene transfer pipettes. Built-in pipette bulb eliminates need for plugging, packaging and autoclaving.
Thermo Scientific™ Disposable Transfer Pipets Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Chemically inert, polyethylene pipets.
Samco™ Transfer Pipettes Available on GSA/VA Contract for Federal Government customers only.
Prevent cross-contamination with these general purpose transfer pipettes, which have a built-in bulb that eliminates the need for plugging.
Thermo Scientific™ Samco™ Special Purpose Transfer Pipettes
Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Special Purpose Transfer Pipettes, ideal for delivering drops onto slides or tiles.
Thermo Scientific™ Samco™ Extra Long Transfer Pipettes Available on GSA/VA Contract for Federal Government customers only.
Perform safe and easy sampling from large containers using Thermo Scientific™ Samco™ Extra-Long Transfer Pipettes, perfect for tissue culture applications.
Thermo Scientific™ Samco™ Fine Tip Transfer Pipettes Available on GSA/VA Contract for Federal Government customers only.
Perform precise dispensing of small drops into small-volume containers using Thermo Scientific™ Samco™ Fine-Tip Transfer Pipettes.
Thermo Scientific™ Samco™ Narrow Stem Transfer Pipettes Available on GSA/VA Contract for Federal Government customers only.
Perform blood bank applications such as sedimentation procedures with this pipette, which feature a flexible stem that bends to sample from narrow or small tubes.
Fisherbrand™ Disposable Soda Lime Glass Pipets Products for Science Education
Ideal for transferring samples to sample cups and for drop counting.
Fisherbrand™ Pasteur Pipets Available on GSA/VA Contract for Federal Government customers only.
Being packaged in strong corrugated material provides maximum protection from external shock for Fisherbrand™ Pasteur Pipets. Each pack also contains a foam pad to protect the pipet tips.
Fisherbrand™ Disposable Graduated Transfer Pipettes Available on GSA/VA Contract for Federal Government customers only.
Ideal for hematology, urinalysis, chemistry, and blood banking — anywhere there is a need for quick, safe transfer of fluids
Gosselin™ Sterile Polypropylene Straw Pipets Available on GSA/VA Contract for Federal Government customers only.
Decrease cross contamination risks using disposable pipets
Kimble™ Kontes™ NMR Pipets Available on GSA/VA Contract for Federal Government customers only.
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
Bel-Art™ SP Scienceware™ Bellows Samplers Available on GSA/VA Contract for Federal Government customers only.
Provide quick and easy sampling with one-handed operation
Kimble™ Disposable Pasteur Pipets Available on GSA/VA Contract for Federal Government customers only.
Borosilicate glass; For one-time use
Bel-Art™ SP Scienceware™ Transpette™ Eight Channel Transfer Pipettor Available on GSA/VA Contract for Federal Government customers only.
For washing plates, feeding, and supernatant aspiration from microwell plates
Corning™ Disposable Glass Pasteur Pipets Available on GSA/VA Contract for Federal Government customers only.
Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.
Bel-Art™ SP Scienceware™ Transpette™ Eight-Channel Disposable Transfer Pipettor Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Perform plate washing, media change and supernatant collection without the use of pipettors and pipette tips with this inexpensive, eight-channel transfer pipettor
Fisherbrand™ Glass Droppers
Straight design glass droppers used to transfer liquid
Fisherbrand™ Disposable Soda-Lime Glass Pasteur Pipets Available on GSA/VA Contract for Federal Government customers only.
Ideal for transferring samples to sample cups and for drop counting
Bel-Art™ SP Scienceware™ Sampler Syringes Available on GSA/VA Contract for Federal Government customers only.
Polypropylene tube can be placed into hot liquids up to 80°C without fear of damage
EISCO Dropping Pipette Products for Science Education
For measuring and transferring liquids. EISCO Pipette Dropping for use in laboratories.
Fisherbrand™ Disposable Flint-Glass Pasteur Pipets in Stand-Up Case Available on GSA/VA Contract for Federal Government customers only.
Tip openings range from 0.03 to 0.06 in. (0.80 to 1.40mm)
  spinner