Prepared Media Plates
LaMotte™ BioPaddles™ Dual-Agar Paddles for Environmental Education Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Contain microbe-specific media enclosed in a sterile vial.
LaMotte™ BioPaddle™ Total Coliform Activity Products for Science Education
For fast and reliable cultivation and enumeration of coliform bacteria.
Carolina™ Nutrient Agar 1.5%, Prepared Media Plates 100 x 15mm Products for Science Education
Carefully tested for sterility and to verify their ability to support the characteristic growth of appropriate organisms.
Carolina™ Nutrient Agar, Prepared Media Plates 100 x 15mm Products for Science Education
An industry standard for nearly a century, this excellent prepared media supports a wide range of microorganisms
Carolina™ MacConkey Agar, Prepared Media Plates 100 x 15mm Products for Science Education
Prepared Media Plates are carefully tested for sterility and to verify their ability to support the characteristic growth of appropriate organisms.
Carolina™ Tryptic Soy Agar, Prepared Media Plates 100 x 15mm Products for Science Education
Carefully tested for sterility and to verify their ability to support the characteristic growth of appropriate organisms.
Carolina™ Mannitol Salt Agar, Prepared Media Plates 100 x 15mm Products for Science Education
Prepared Media Plates are carefully tested for sterility and to verify their ability to support the characteristic growth of appropriate organisms.
Thermo Scientific™ Sabouraud Dextrose Agar Available on GSA/VA Contract for Federal Government customers only.
Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, on a ready-to-use prepared media plate.
Thermo Scientific™ Remel™ Anaerobic Reducible Neomycin Blood Agar
Columbia Blood Agar base used for selective isolation of anaerobic microorganisms such as Bacteroides and Clostridium spp.
Thermo Scientific™ Remel™ Anaerobic Reducible Blood Agar Available on GSA/VA Contract for Federal Government customers only.
Blood agar (Columbia Blood Agar base) is used for the primary cultivation and isolation of fastidious anaerobic microorganisms.
Thermo Scientific™ Remel™ Blood Agar (TSA w/5% Sheep Blood)/ SXT Blood Agar
Used for the isolation and/or differentiation of microorganisms or specimens
Thermo Scientific™ Remel™ Cornmeal Agar w/Polysorbate 80 Available on GSA/VA Contract for Federal Government customers only.
General purpose medium for the cultivation of fungi and the demonstration of chlamydospore production by Candida albicans.
Thermo Scientific™ Remel Slide Culture Plate, Potato Dextrose Agar Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Slide Culture Plate, Potato Dextrose Agar is for morphological and microscopic identification of fungi.
Thermo Scientific™ Remel™ Blood Agar/ Blood Agar (TSA w/5% Sheep Blood) Available on GSA/VA Contract for Federal Government customers only.
Used for the isolation and/or differentiation of microorganisms or specimens
Thermo Scientific™ Remel™ Contact I Mycological Agar w/Lecithin, Polysorbate 80
Thermo Scientific™ Remel Contact I Mycological Agar w/Lecithin, Polysorbate 80 has a low pH for cultivation and isolation of yeast and fungi.
Thermo Scientific™ Remel™ Biplates: Anaerobic Reducible through Levine Agars Available on GSA/VA Contract for Federal Government customers only.
Perform screening and isolation in the same 12 x 85mm dish, divided into two sections to hold either the same media on both sides or two media, one on each side.
Thermo Scientific™ Remel™ Potato Flake Agar Slide Culture Plate Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Potato Flake Agar Slide Culture Plate is used for morphological and microscopic identification of fungi.
Thermo Scientific™ Remel™ Anaerobic KV Blood Agar Available on GSA/VA Contract for Federal Government customers only.
Blood agar (CDC formulation) with Kanamycin and vancomycin inhibit gram-positive microorganisms and facultative anaerobic bacteria, and select for gram-negative bacilli.
Thermo Scientific™ Remel™ Sabouraud Dextrose Agar, Emmons w/Chloramphenicol, Cycloheximide Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Sabouraud Dextrose Agar, Emmons with Chloramphenicol, Cycloheximide is used for the selective isolation of yeast and fungi.
Thermo Scientific™ Mueller Hinton Agar Available on GSA/VA Contract for Federal Government customers only.
Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium which may be used in internationally recognized standard procedures.
Thermo Scientific™ Blood Agar (TSA with Sheep Blood) Medium Available on GSA/VA Contract for Federal Government customers only.
Grow a wide variety of microorganisms on this general purpose non-selective medium that includes sheep blood to promote haemolysis.
Thermo Scientific™ Remel™ SABHI™ Agar w/Chloramphenicol, Cycloheximide Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel SABHI Agar w/Chloramphenicol, Cycloheximide is for the cultivation, isolation of pathogenic fungi.
Thermo Scientific™ Remel™ Contact I Rose Bengal Agar
Thermo Scientific™ Remel Contact I Rose Bengal Agar is used for selective isolation and enumeration of yeast, and fungi from environmental samples and food.
Thermo Scientific™ Remel™ BHI Agar w/Vancomycin Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel BHI Agar w/Vancomycin (6μg/mL) is a screening test for detection of vancomycin resistance in enterococci.
Thermo Scientific™ Remel™ Brucella Agar w/5% Sheep Blood, Trimethoprim, Vancomycin, Polymyxin B
For primary cultivation and isolation of Campylobacter jejuni from fecal specimens or rectal swabs.
Thermo Scientific™ Remel™ Settling Plate, Sterile, Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar w/Lecithin and Polysorbate 80 facilitates cultivation of yeast and fungi.
Thermo Scientific™ Remel™ XLD Agar (Xylose, Lysine, Desoxycholate) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel XLD Agar (Xylose, Lysine, Desoxycholate) is for the selective and differential isolation of gram-negative enteric pathogens.
  spinner