Gas Bubblers and Receivers
Kimble™ Kontes™ Gas Bubbler
Used with mineral oil to observe gaseous addition to reactions
Kimble Chase Kontes™ Gas-Sampling Midget Impinger Available on GSA/VA Contract for Federal Government customers only.
Smaller version of the Greenburg-Smith impinger
Kemtech America™ Synthware™ Mineral Oil Bubbler
Used to monitor gas flow to reactions while providing a vent to the atmosphere.
  spinner