Allihn Condensers
PYREX™ Allihn Condensers with standard taper Joints Available on GSA/VA Contract for Federal Government customers only.
Suited for use in vacuum distillation
Kimble™ Kontes™ Allihn Condensers with Straight Outlet Tube
For refluxing applications including Soxhlet extractions
Wheaton™ Allihn Condensers For Soxhlet Extraction Apparatus Available on GSA/VA Contract for Federal Government customers only.
Used for continuous extraction of analytes from a solid into an organic solvent
PYREX™ Replacement Allihn Condenser Available on GSA/VA Contract for Federal Government customers only.
Replacement Allihn condenser with bulbs
Kimble™ Kontes™ Allihn Condensers with Full Length Joints
Allihn reflux condenser with a standard taper drip joint and a standard taper outer
Kemtech America™ Synthware™ Allihn Condenser
With joints at the top and the bottom
  spinner