Glass Specialty Rotary Traps
Kimble™ Kontes™ Rotary Evaporator Traps With Inner Vapor Tube at Top
Design allows for expanded volume for mixtures that have a tendency to foam excessively
PYREX™ Evaporator Traps Available on GSA/VA Contract for Federal Government customers only.
Used when foam is likely to be created and drawn into the condenser of rotary evaporator
Kimble™ Kontes™ Rotary Evaporator Traps With Inner Vapor Tube at Bottom Available on GSA/VA Contract for Federal Government customers only.
Use when foam is likely to be created and drawn into the condenser
Fisherbrand™ Rotary Evaporator Trap with Drain Holes
Use where foaming is likely to occur and be drawn into the condenser.
  spinner