Glass Specialty Tubes
Fisherbrand™ Microhematocrit Capillary Tubes Available on GSA/VA Contract for Federal Government customers only.
Low-taper wall construction guarantees error less than 1% absolute, 0.4 hematocrit, relative
PYREX™ Tube Adapters with standard taper Joints at 105° Angle Available on GSA/VA Contract for Federal Government customers only.
Two-way tube with inner and outer standard taper 24/40 joint connections at 105° angle
Separation Technology, Inc. ClearCRIT™ Plastic Capillary Tubes
For use with standard microhematocrit centrifuges, including HemataSTAT
PYREX™ Tube Adapters with Two standard taper Joints and Thermometer Opening Available on GSA/VA Contract for Federal Government customers only.
Connecting three-way tube with side arm placed at 75° angle from lower joint
Separation Technology, Inc. ClearCRIT™ Capillary Tubes
Mylar™ wrapped glass tubes offer an added layer of safety without compromising accuracy
PYREX™ Vacuum Distilling Adapter Tube Available on GSA/VA Contract for Federal Government customers only.
Converts flasks with standard taper necks to vacuum-type standard taper receivers
PYREX™ Tube Adapters with standard taper Joints at 75° Angle Available on GSA/VA Contract for Federal Government customers only.
Two-way tube with standard taper 24/40 inner joints at both ends
PYREX™ Tube Adapters with Outer standard taper Joint Available on GSA/VA Contract for Federal Government customers only.
The lower end is cut off at an angle for use as receiver adapter.
PYREX™ Connecting 3-Way Claisen Adapters with Parallel Side Arms Available on GSA/VA Contract for Federal Government customers only.
Three-way connecting tube with two outer and one inner standard taper 24/40 joints
PYREX™ Vacuum Connecting Adapter Tubes Available on GSA/VA Contract for Federal Government customers only.
105° angle connector with 10mm O.D. tubulation for attaching to vacuum pump
PYREX™ Tube Adapters with 75° Angle and Three standard taper Joints Available on GSA/VA Contract for Federal Government customers only.
Three-way connecting tube with side arm placed at 75° angle from lower joint
Kimble™ Kontes™ Schlenk Vacuum Tube with HI-VAC Valve Available on GSA/VA Contract for Federal Government customers only.
Suitable for use with K216050 or K216060 Manifolds
Fisherbrand™ Plastic Microhematocrit Tubes Available on GSA/VA Contract for Federal Government customers only.
Clarity and performance of glass—shatterproof safety of plastic
StatSpin™ SafeCrit™ Plastic Microhematocrit Tubes Available on GSA/VA Contract for Federal Government customers only.
Plastic tubes can be bent or dropped without shattering
Drummond Scientific Hemato-Clad™ Mylar™-Wrapped Hematocrit Tubes Available on GSA/VA Contract for Federal Government customers only.
Combine the precision of glass with the safety of Mylar wrapping
  spinner