Glass Tissue Grinders
Kimble™ Kontes™ Pellet Pestle™ Available on GSA/VA Contract for Federal Government customers only.
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Wheaton™ Potter-Elvehjem Tissue Grinders With Radial Serrations Available on GSA/VA Contract for Federal Government customers only.
Serrated PTFE pestle disperses homogenate into the mortar cylinder more efficiently
Wheaton™ Tenbroeck Tissue Grinder
With expanded reservoir and pouring lip
Wheaton™ Potter-Elvehjem Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Choose grinders with or without plastic safety coating
Wheaton™ Dounce Tissue Grinders
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
Kimble™ Kontes™ Potter-Elvehjem Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
With serrated PTFE pestle and graduated tube
Kimble™ Kontes™ Micro Duall™ Tissue Grinder
Heavy-walled construction minimizes tube breakage
Kimble™ Kontes™ Pellet Pestle™ Cordless Motor Available on GSA/VA Contract for Federal Government customers only.
Produces more complete disaggregation than manual mixing
Kimble™ Kontes™ Dounce Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Designed for cellular work in which the nucleus remains intact after homogenization
PYREX™ PTFE Pestle Tissue Grinder with Steel Shaft Available on GSA/VA Contract for Federal Government customers only.
Recommended for soft tissues found in the liver and brain
PYREX™ Glass Pestle Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Consist of glass tube and pestle with clearance of 0.15mm
Wheaton™ Tapered Tissue Grinders With PTFE Pestles Available on GSA/VA Contract for Federal Government customers only.
Tapered tissue grinder with a PTFE pestle for soft tissues
Wheaton™ Grinding Chamber Replacement Parts For Tapered Tissue Grinders With PTFE Pestles Available on GSA/VA Contract for Federal Government customers only.
Tapered tissue grinders replacement grinding chamber for fluoropolymer resin pestle
Fisherbrand™ Pellet Pestle™ Cordless Motor Adapter Available on GSA/VA Contract for Federal Government customers only.
Adapter for Fisherbrand Pellet Pestle Cordless Motor
PYREX™ Potter-Elvehjem Tissue Grinder with PTFE Pestle Available on GSA/VA Contract for Federal Government customers only.
Used in preparing small samples for pathological and biochemical studies
PYREX™ Dounce Tissue Grinder with two Pestles Available on GSA/VA Contract for Federal Government customers only.
Two pestles dissociate cells into fine particles with minimal damage to cell nuclei
Kimble™ Kontes™ Pellet Pestle™ Cordless Motor: Replacement Motor Adapter
Replacement parts for Kontes Pellet Pestle Cordless Motor
PYREX™ Tenbroeck Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Designed for hand operation
Kimble™ Kontes™ All-Glass Potter-Elvehjem Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Designed for motor drive using 788000 Stirrer Adapter
Fisherbrand™ Pellet Pestles™ Available on GSA/VA Contract for Federal Government customers only.
Resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
  spinner