Glass Centrifuge Tubes
Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 12.5mL Capacity Available on GSA/VA Contract for Federal Government customers only.
Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements.
Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes, 10mL Available on GSA/VA Contract for Federal Government customers only.
Use these tubes for centrifugation up to 2980 RCF. KIMAX™ Conical-Bottom Glass Centrifuge Tubes are plastic shrink-wrapped in modular trays for cleanliness and safety. Manufactured from 51 expansion, low extractable borosilicate glass.
Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 2mL Capacity
Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements.
Corning™ Conical-Bottom Glass Centrifuge Tubes: 3mL Capacity Available on GSA/VA Contract for Federal Government customers only.
Nongraduated with beaded rim; no closure.
Kimble™ Kontes™ KIMAX™ Reusable High Strength Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Chemically strengthened to achieve a greater mechanical strength
PYREX™ Centrifuge Tubes Products for Science Education
For microchemical applications.
Kimble™ Kontes™ KIMAX™ Reusable Threaded Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Heavy wall withstands high centrifuge speeds
Bel-Art™ SP Scienceware™ 50mL Conical Tube Holders Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Leave both hands free for pipetting and other critical tasks.
Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 100mL Capacity
Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements.
  spinner