Glass Test Tubes
PYREX™ VISTA™ Test Tubes with Beaded Rim Products for Science Education
Functionality, economy, and quality.
Kimble™ KIMAX™ Reusable Tubes with Rubber-Lined Screw Cap Available on GSA/VA Contract for Federal Government customers only.
Black phenolic cap with inert rubber liner
PYREX™ Reusable Borosilicate Glass Tubes with Plain End Available on GSA/VA Contract for Federal Government customers only.
All sizes except 6, 22, and 25mm O.D. have marking spot
PYREX™ Ignition Tubes Products for Science Education
An excellent choice for classroom applications.
PYREX™ Reusable Glass Tubes with Rubber-Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Deep-form phenolic caps facilitate handling and sealing after autoclaving
Fisherbrand™ Reusable Glass Tubes with Phenolic Screw Caps Available on GSA/VA Contract for Federal Government customers only.
Phenolic screw cap with white rubber liner
PYREX™ Test Tubes Products for Science Education
Beaded rim for durability.
Kimble™ Amber Plain Disposable Borosilicate Glass Tubes
Produced from amber borosilicate glass to give superior protection for light sensitive applications.
Kimble™ KIMAX™ Disposable Glass Tubes Available on GSA/VA Contract for Federal Government customers only.
Use these glass tubes for a variety of applications and cell washing procedures. Kimble™ KIMAX™ Disposable Glass Tubes are made from 51 expansion borosilicate glass, which offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products.
PYREX™ VISTA™ Reusable Test Tubes with Beaded Rim Available on GSA/VA Contract for Federal Government customers only.
Functionality, economy, and quality
PYREX™ Screw Cap Culture Tubes with PTFE Lined Phenolic Caps Available on GSA/VA Contract for Federal Government customers only.
PTFE interface is firmly bonded to glued-in rubber liner
Kimble™ ValueWare™ Test Tubes Products for Science Education
Made from Kimax™ KG-33 glass tubing.
Fisherbrand™ Disposable Borosilicate Glass Tubes with Polypropylene Screw Cap Available on GSA/VA Contract for Federal Government customers only.
Provides chemical stability and biological inertness of hard, borosilicate glass — at price of disposable tubes
Fisherbrand™ Disposable Flint Glass Tubes with Plain End Available on GSA/VA Contract for Federal Government customers only.
Specially annealed to reduce breakage along stress point
PYREX™ Reusable Heavy-Wall Ignition Tubes Available on GSA/VA Contract for Federal Government customers only.
For high-temperature work in the classroom
Kimble™ KIMAX™ Reusable Tubes with PTFE-Faced Rubber-Lined Screw Cap Available on GSA/VA Contract for Federal Government customers only.
Black phenolic cap with PTFE-faced rubber liner
PYREX™ Reusable Test Tubes with Beaded Rim Available on GSA/VA Contract for Federal Government customers only.
Sizes from 10 to 20mm O.D. have marking spot
3B Scientific™ Capillary Tubes Apparatus Products for Science Education
Demonstrates relationship between the bore of the tube and capillary pressure.
  spinner