NMR Tubes
Kimble™ Kontes™ Brand 3mm NMR Tubes Available on GSA/VA Contract for Federal Government customers only.
Each tube meets rigid specifications for wall thickness, variation and camber so as to assure precise measurements and reproducibility.
Kimble™ Kontes™ NMR Seal Off NMR Tubes 5mm
Includes polyethylene closure
Kimble™ Kontes™ Brand 5mm High Grade NMR Sample Tubes Available on GSA/VA Contract for Federal Government customers only.
Each tube meets rigid specifications for wall thickness, variation and camber so as to assure precise measurements and reproducibility.
Wilmad™ SP Scienceware™ 10mm O.D. Thin Walled Quartz NMR Tube Available on GSA/VA Contract for Federal Government customers only.
Wilmad Precision NMR Quartz Tubes have an extremely low thermal expansion rate and high tensile strength, which is 14 times more robust during the cooling/heating process than Type 1 Class A glass tubes
Corning™ Precision Economy NMR Tubes Available on GSA/VA Contract for Federal Government customers only.
Precision machined for exact wall thickness, perfect straightness and centerline symmetry
PYREX™ 5mm O.D. NMR Sample Tubes Available on GSA/VA Contract for Federal Government customers only.
Precision NMR tubes made of Pyrex borosilicate glass
Kimble™ Kontes™ KIMAX™ NMR Borosilicate Glass 5mm Tube
Ideal for sealing directly to vacuum manifolds, joints or valves
Kimble™ Kontes™ KIMAX™ Borosilicate Glass Thrift N-51A 5mm NMR Tubes Available on GSA/VA Contract for Federal Government customers only.
Used to contain samples in nuclear magnetic resonance spectroscopy
Kimble™ Kontes™ Brand 5mm Disposable Grade NMR Sample Tubes Available on GSA/VA Contract for Federal Government customers only.
For instruments with field strength of approximately 60MHz
Wilmad™ SP Sciencware™ 5mm NMR/EPR Tube Label
Fits the circumference of the NMR tube precisely with no overlapping to guarantee sample tube's symmetry in spinning experiment. Bel-Art SP Scienceware™ Wilmad™ 5mm NMR/EPR Tube Label is a handy alternative to marking your NMR or EPR sample tubes.
Kimble™ Kontes™ Brand Valved 5mm O.D. NMR Tubes Available on GSA/VA Contract for Federal Government customers only.
Ideal for handling volatile, air-sensitive and toxic samples
Corning™ Precision Isostatic Pressure Strength NMR Tubes Available on GSA/VA Contract for Federal Government customers only.
Precision machined for exact wall thickness, perfect straightness and centerline symmetry
Kimble™ Kontes™ Threaded NMR Tubes with Cap
For air-sensitive samples and reaction studies where compounds are added via syringe through septum cap
Kimble™ Kontes™ KIMAX™ N-51A Borosilicate Glass Flat Bottom NMR Tubes Available on GSA/VA Contract for Federal Government customers only.
Ideal for use in high resolution NMR spectroscopy
  spinner