Glass Tubing Connectors and Accessories
Kimble™ Kontes™ Reducing Adapters
For connecting dissimilar standard taper joints
Kimble™ Kontes™ Gas Inlet Adapters
Allows access to the bottom of flasks/tubes
Kimble™ Kontes™ Hose Inlet Adapter with Vacuum Take Off
Vertical adapter with vacuum take-off and drip tip
Fisherbrand™ Thin PTFE Sleeves
Elongated cones of microthin (0.05mm) polytetrafluoroethylene film tapered to fit snugly around glass joints
Kimble™ Kontes™ Hose Inlet Adapters
For use as a vent or for the introduction of pressure/vacuum
Corning™ Enlarging Adapters
Pyrex™ glass adapter with top outer joint; larger bottom inner joint.
Kimble™ Kontes™ Bevel-Seal™ Inlet Adapter
For use with pipets, syringes, or other small items with outer diameters from 0.06 to 0.62 in. (2 to 17mm)
Fisherbrand™ Pure Low Density PTFE Thread Seal Tape
Prevents leaks to ensure a proper seal when connecting items, such as a pressure gauge to an adaptor
PYREX™ Enlarging Glass Joint Adapters Available on GSA/VA Contract for Federal Government customers only.
For attaching apparatus with one standard taper joint size to equipment having larger size standard taper joint
Kimble™ Kontes™ Vacuum Filtration Adapters
Use for reduced pressure filtration with plain stem Buchner funnels.
Kimble™ Kontes™ Oft Adapter
Bevel-Seal; Offset; 10° angle for multi-neck flasks
PYREX™ Reducing Glass Joint Adapters Available on GSA/VA Contract for Federal Government customers only.
For attaching apparatus with one standard taper joint size to equipment having a different size standard taper joint
PYREX™ Bleed Tubes With Capillary Tip Available on GSA/VA Contract for Federal Government customers only.
For bleeding gases, steam or liquids
Kimble™ Kontes™ Bevel-Seal Vacuum Inlet Adapters
For use with an O.D. from 2 to 17mm
  spinner