Glass Joint Adapters
PYREX™ Capillary Tubing Available on GSA/VA Contract for Federal Government customers only.
Low-expansion, borosilicate glass tubing
GE Healthcare Tubing Accessory for Hoefer™ GD 2000 Gel Drying System Available on GSA/VA Contract for Federal Government customers only.
VT3 Vacuum Tubing for Hoefer™ GD 2000 Gel Drying System
Pyrex™ Brand Standard Wall Tubing
Low coefficient of expansion, great mechanical strength
GSC International Flint Glass Tubing Products for Science Education
Useful in science and chemistry applications.
Glass Tubing Products for Science Education
Laboratory tubing with standard wall.
Kemtech America™ Synthware™ Inlet Adapters with Fritted Disc
With extra coarse porosity fritted disc to prevent solid from being introduced into reaction system during liquid transfer
Kemtech America™ Synthware™ Twin Connecting Hose Adapters
Two hose connections 180° from one another to permit a flow of inert gas over the apparatus
Kemtech America Synthware™ Vacuum Transfer Adapter with PTFE Stopcocks
Adapter with 2mm stopcock used for the transfer of sensitive and hazardous solutions
Kemtech America Synthware™ Claisen Adapter with 45° Side Arm
Claisen adapter with an additional standard taper outer joint at a 45 deg. angle to the vertical outer joint
Kemtech America™ Synthware™ Flow Control Adapters with 1:5 Teflon™ Stopcock
Used to control the flow of gasses or liquids into reaction systems
Kemtech America Synthware™ Connecting Adapter
Three-arm glass joint adapters for the assembly of three vessels
Synthware™ Adapter with Mineral Oil Bubbler
Allows in-line connection and purging of apparatus
Kemtech America™ Synthware™ Distilling Adapter
Useful in sub-boiling separation of mixtures having volatile components
Kemtech America™ Synthware™ Enlarging Adapters
With standard taper joints at top and bottom
Kemtech America™ Synthware™ Flow Control Adapters with PTFE Stopcock
Used to regulate the flow of gases or liquids into reaction systems
Kemtech America™ Synthware™ Distillation Connecting Adapters, Angled at 75°
For use as a component in atmospheric or vacuum distillations
Kemtech America Synthware™ Distillation Adapter, 75°
Distillation adapter with standard taper inner joints on each end at a 75 deg. angle
Kemtech America™ Synthware™ Vacuum Adapters with Outer Joint
Ideal for use as a vent or for the introduction of pressure or vacuum
Kemtech America™ Synthware™ Inlet or Thermometer Adapters
With a compression cap and Viton O-ring to form a vacuum tight seal
Kemtech America™ Synthware™ Flushing Adapters with PTFE Stopcock
Allows systems to be flushed with an inert gas while sampling directly through the bore of the T-bore stopcock
Kemtech America™ Synthware™ Vacuum or Argon Adapters
With hose connection on the side and male joint
Kemtech America™ Synthware™ U-Shape Adapter
With standard taper inner joints at both ends
Kemtech America™ Synthware™ Inlet/Thermometer Adapter, Microscale
Inlet or thermometer adapter with a compression cap and a Viton® O-ring to form a tight seal. Cap accommodates a 5.0-7.5mm O.D. plain stem thermometer. Supplied with one Nylon® cap and one Viton® O-ring. For microscale applications
Kemtech America™ Synthware™ Reducing Adapters
With standard taper joints at top and bottom
Kemtech America™ Synthware™ Adapters with Ball Socket
With top spherical socket joint and lower standard taper inner joint
Kemtech America™ Synthware™ Bushing Adapters
For compact assemblies of dissimilar joints
Kemtech America™ Synthware™ Drying Tube Adapters, Angled at 75°
Angled at 75° to avoid desiccant contact with reactions
  spinner