Glass Joint Sleeves
Kimble™ Kontes™ Bevel-Seal™ Inlet Adapter
For use with pipets, syringes, or other small items with outer diameters from 0.06 to 0.62 in. (2 to 17mm)
Kimble™ Kontes™ Reducing Adapters
For connecting dissimilar standard taper joints
PYREX™ Bushing Type Reducing Glass Joint Adapters Available on GSA/VA Contract for Federal Government customers only.
With standard taper inner and outer joints for changing standard taper joint sizes
PYREX™ Reducing Glass Joint Adapters Available on GSA/VA Contract for Federal Government customers only.
For attaching apparatus with one standard taper joint size to equipment having a different size standard taper joint
Kimble™ Kontes™ Hose Inlet Adapter with Vacuum Take Off
Vertical adapter with vacuum take-off and drip tip
PYREX™ Enlarging Glass Joint Adapters Available on GSA/VA Contract for Federal Government customers only.
For attaching apparatus with one standard taper joint size to equipment having larger size standard taper joint
  spinner