Glass Scintillation Vials
Wheaton™ Glass 20mL Scintillation Vials: Polypropylene Caps
Maintain integrity of seal with variety of cap liners. Wheaton Science Products™ Glass 20mL Scintillation Vials: Polypropylene Caps are Chemically inert, suitable for strong solvents and high-purity chemicals.
Kimble™ 20mL Glass Screw-Thread Scintillation Vials
Manufactured from clear KG-33 borosilicate glass
Fisherbrand™ Borosilicate Glass Scintillation Vials, with white polypropylene caps, 20mL
Use these general purpose vials with all common counters. Fisherbrand™ 20mL Borosilicate Glass Scintillation Vials with white polypropylene caps feature potassium-free glass composition which provides consistently low activity counts and excellent light transmission.
Kimble™ 7mL Solvent-Saver™ Scintillation Vials
Use same volume of radioactive sample as 20mL vials for savings in solvent and solvent disposal costs
Wheaton™ Glass 20mL Scintillation Vials and Urea Caps
Maintains integrity of seal with variety of cap liners. Wheaton Science Products™ Glass 20mL Scintillation Vials and Urea Caps are suitable for strong solvents and high-purity chemicals.
Fisherbrand™ 7mL Borosilicate Glass Scintillation Vial
Smaller capacity vial saves in solvent use and disposal
  spinner