Cell Strainers
Fisherbrand™ Cell Strainers
Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.
Corning™ Sterile Cell Strainers Available on GSA/VA Contract for Federal Government customers only.
Isolates primary cells rapidly to consistently obtain a uniform single-cell suspension from tissues. Corning™ Sterile Cell Strainers are ideal for preparation of flow cytometry samples and are used with stem cells and primary cells.
Bel-Art™ SP Scienceware™ Flowmi™ Cell Strainers for 1000ul Pipette Tips Available on GSA/VA Contract for Federal Government customers only.
Provides fast and efficient filtering of small volume samples (up to 1000µl) prior to FLOW or FACS analysis (patent pending.) Bel Art™ SP Scienceware™ Flowmi™ Cell Strainers for 1000ul Pipette Tips are sterile and come in two porosity sizes.
Corning™ Falcon™ Test Tube with Cell Strainer Snap Cap Available on GSA/VA Contract for Federal Government customers only.
Designed for flow cytometry applications. Corning™ Falcon™ Test Tube with Cell Strainer Snap Cap offer a convenient solution to prepare laboratory samples.
Falcon™ Cell Strainers Available on GSA/VA Contract for Federal Government customers only.
Rapidly isolates primary cells to consistently obtain a uniform single-cell suspension from tissues. Corning Falcon™ Cell Strainers are easy to use and ideal for use with stem cells and primary cells.
  spinner