Plastic Beakers
EISCO Polypropylene Beakers with Spout Products for Science Education
Excellent clarity and very good chemical resistance.
Bel-Art™ SP Scienceware™ Griffin Low Form Polypropylene beakers Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Rugged beakers have permanent, raised numeric graduations and no-drip pour spout for easy pouring.
Corning™ Reusable Polypropylene Low-Form Beakers Products for Science Education
Translucent polypropylene beakers printed with blue graduations.
Corning™ Reusable Plastic Beakers with Handle and Spout Products for Science Education
Translucent polypropylene beakers printed with blue graduations.
3B Scientific™ Low Form Beakers Products for Science Education
Low form beakers are made up of transparent TPX material.
United Scientific Supplies Stackable Beakers Products for Science Education
Highly flexible polypropylene beakers.
United Scientific Supplies Polypropylene Tri-Corner Beakers Products for Science Education
Tri-corner design that provides three dripless pouring spouts.
United Scientific Supplies Plastic Beakers Products for Science Education
Excellent clarity and very good chemical resistance.
United Scientific Supplies Disposable Plastic Beaker Products for Science Education
Beakers, molded in polypropylene, have excellent clarity and very good chemical resistance.
Dynalon™ Polymethylpentene Beakers Products for Science Education
A safe alternative to glass in the classroom lab.
GSC International Polypropylene Beaker Jug Products for Science Education
Includes a handle and pour spout for ease of use. The GSC International Polypropylene Beaker Jug is graduated and available in several sizes.
Dynalon™ PTFE Heatable Beaker Products for Science Education
250mL PTFE fluoropolymer heatable beaker.
Dynalon™ Plastic Beaker with Handle Products for Science Education
Our handles make your handling of solutions easier and safer.
Dynalon™ Polypropylene Beakers Products for Science Education
Chemicals can't wear this beaker down
GSC International Polypropylene Graduated Cylinders, 1000mL Products for Science Education
Feature raised graduations for easy reading. GSC International Polypropylene Graduated Cylinders incorporate a hex base for greater stability, and are autoclavable.
GSC International Euro Style Polypropylene Beakers Products for Science Education
Incorporate raised outer graduations for ease of reading. GSC International Euro Style Polypropylene Beakers are tough, chemical resistant, and autoclavable. They are great additions to laboratory or classroom settings.
Thermo Scientific™ Nalgene™ PMP Graduated Plastic Beakers with Handles Available on GSA/VA Contract for Federal Government customers only.
Ensure resistance against acids, bases, aliphatic alcohols, aldehydes and esters with Thermo Scientific™ Nalgene™ PMP Graduated Beakers.
Thermo Scientific™ Nalgene™ PFA Graduated Plastic Beakers with Handles Available on GSA/VA Contract for Federal Government customers only.
Conduct high-purity applications that use strong oxidizing agents, acids, hydrocarbons and ketones with Thermo Scientific™ Nalgene™ PFA Graduated Beakers with Handles.
Thermo Scientific™ Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers Available on GSA/VA Contract for Federal Government customers only.
Carry out precise and efficient pouring with Thermo Scientific™ Nalgene™ Polypropylene Griffin Low-Form Beakers, featuring a single-spout design.
Thermo Scientific™ Nalgene™ PMP Griffin Low-Form Plastic Beakers Available on GSA/VA Contract for Federal Government customers only.
Continuously use Thermo Scientific™ Nalgene™ PMP Griffin Low-Form Beakers at temperatures of 150°C with intermittent service to 175°C.
Thermo Scientific™ Nalgene™ Griffin Low-Form PFA Plastic Beakers Available on GSA/VA Contract for Federal Government customers only.
Handle temperatures from -270° to +250°C using sturdy and tough Thermo Scientific™ Nalgene™ Griffin Low-Form PFA Beakers, ideal for continual lab usage.
  spinner