Plastic Environmental Sampling Bottles
Thermo Scientific™ LDPE Cubitainers™ with LDPE Foam-lined Closure
Light weight,space-efficient solution for sampling in remote locations. Available certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride.
Thermo Scientific™ Wide-Mouth HDPE Packers with Closure
Recommended for collecting water, leachate, and other liquid, semi-liquid, or viscous samples to be analyzed for inorganic trace analytes.
Thermo Scientific™ Wide-Mouth HDPE Oblong with Lined Closure
Save valuable space and stabilize samples within the shipping container. Certified to meet EPA Performance Based Specifications for metals, cyanide and fluoride analyses.
Thermo Scientific™ Nalgene™ Certified Wide-Mouth Amber HDPE Bottle with Closure
Recommended for critical projects where sample containment and transport is of elevated concern, featuring durable construction that withstands the rigors of field operations.
Thermo Scientific™ Nalgene™ Certified Wide-Mouth HDPE Bottle with Polypropylene Screw Closure
Recommended for critical projects where sample containment and transport is of elevated concern, featuring durable construction that withstands the rigors of field operations.
Thermo Scientific™ Wide-Mouth HDPE Packers without Caps: Bulk Pack
HDPE Wide-Mouth Packers are general-purpose containers, bulk and unassembled. Unprocessed, ready for your own cleaning procedure and assembly with caps purchased separately.
Thermo Scientific™ Particle Certified HDPE Containers
Protect the quality of your products or lab samples with these reliable particle-certified HDPE containers,certified to contain < 20 particles per milliliter = 0.3 microns.
Thermo Scientific™ Bulk Pack HDPE Cylinder Rounds without Caps
HDPE Cylinder Round is a general-purpose container. Bulk and unassembled. Unprocessed, ready for your own cleaning procedure and assembly with caps purchased separately.
Thermo Scientific™ HDPE Cylinder Round Bottles
Collect water samples and other low-viscosity liquids to be analyzed for inorganic trace analytes with the original EPA-specified bottle for liquid sampling.
Thermo Scientific™ Nalgene™ Certified Narrow-Mouth HDPE Bottle with Polypropylene Screw Closure
Recommended for water and other environmental liquid sample collections, especially for critical projects where sample containment and transport is of elevated concern.
Thermo Scientific™ Nalgene™ Wide-Mouth Teflon™ EP Tox/TCLP Bottle with Closure
Use Thermo Scientific™ Nalgene™ Wide-Mouth Teflon™ EP Tox/TCLP Bottles at high and low temperatures for trace metal analysis and applications with organic solvents.
Thermo Scientific™ Straight-Sided Wide-Mouth HDPE Jar with Closure
Wide-Mouth HDPE Jar allows for easy filling and sample retrieval, and is Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride.
Thermo Scientific™ HDPE Jugs with White LDPE Foam-Lined Polypropylene Screw Closure
Thermo Scientific™ HDPE Jugs meet or exceed EPA performance-based standards for metals, cyanide and fluoride.
Thermo Scientific™ HDPE Narrow-Mouth Boston Round Bottles
Collect water samples and other low viscosity liquids to be analyzed for inorganic trace analytes. These Certified containers, featuring narrow mouth for precision pouring.
Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Sterile Sample Bottle with Closure
Nalgene Wide-Mouth HDPE Sterile Sample Bottles are break-resistant, guaranteed leak-proof and certified sterile for water sample collection for microbiological analyses.
Thermo Scientific™ Security-Snap™ Coliform Polypropylene Water Sample Bottle: Sterile
Security-Snap Sterile Coliform Water Sample Bottle meets all the EPA requirements for microbiology sample containers for the analysis of drinking water.
Thermo Scientific™ Screw-Top Sterile Coliform Water Sample Bottle: Polystyrene, Polyethylene and Polypropylene
Designed for use with EPA compliant total coliform and fecal coliform water analysis using the Colilert analysis method or equivalent.
Fisherbrand™ Pre-Cleaned Straight-Sided Jars
Certified for background contaminants
Wheaton™ Leak-Resistant Wide-Mouth Natural PP Bottles Available on GSA/VA Contract for Federal Government customers only.
With double-seal closure with cap attached
Fisherbrand™ Pre-Cleaned HDPE Wide Mouth Jars
Certified for background contaminants
Wheaton™ Leak-Resistant Narrow-Mouth Natural LDPE Bottles Available on GSA/VA Contract for Federal Government customers only.
With double-seal closure with cap attached
Fisherbrand™ Pre-Cleaned Wide Mouth Leakproof Bottles
Certified for background contaminants
Wheaton™ Leak-Resistant Narrow-Mouth Natural PP Bottles Available on GSA/VA Contract for Federal Government customers only.
With double-seal closure with cap attached
Wheaton™ Leak-Resistant Wide-Mouth HDPE Bottles Available on GSA/VA Contract for Federal Government customers only.
With double-seal closure with cap attached
Fisherbrand™ Pre-Cleaned Cylinders
Certified for background contaminants
Fisherbrand™ Pre-Cleaned Narrow Mouth Leakproof Bottles
Certified for background contaminants
Fisherbrand™ Pre-Cleaned Boston Round Bottles
Certified for background contaminants
Fisherbrand™ Pre-Cleaned Jugs
Certified for background contaminants
Fisherbrand™ Pre-Cleaned Wide Mouth Oblong Bottles
Certified for background contaminants
Wheaton™ Leak-Resistant Wide-Mouth Natural LDPE Bottles Available on GSA/VA Contract for Federal Government customers only.
With double-seal closure with cap attached
  spinner