Plastic Carboys
Thermo Scientific™ Nalgene™ Rectangular HDPE Carboy with Spigot Available on GSA/VA Contract for Federal Government customers only.
Store and dispense media, solutions and sterile water with space-saving Thermo Scientific™ Nalgene™ Rectangular HDPE Carboys.
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboys with Bottom Tubulation Available on GSA/VA Contract for Federal Government customers only.
Ensure more complete drainage with the low outlet of these polypropylene carboys, excellent for distilled water and culture media.
Thermo Scientific™ Nalgene™ Rectangular Autoclavable PPCO Carboys with Spigot Available on GSA/VA Contract for Federal Government customers only.
Store and dispense media, solutions and sterile water. Space-saving design features a wide mouth opening for filling and cleaning and a spigot for easy dispensing.
Thermo Scientific™ Nalgene™ Rectangular Fluorinated HDPE Carboy with Spigot Available on GSA/VA Contract for Federal Government customers only.
Easily fill and clean this carboy. Featuring a wide-mouth opening, space-saving rectangular design and a fluorinated surface to enhance long-term performance.
Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ with Spigot Available on GSA/VA Contract for Federal Government customers only.
Store media, solutions and sterile water in Nalgene Rectangular Clearboys. Featuring a space-saving design and extreme impact resistance. Ideal for cold room use.
Thermo Scientific™ Nalgene™ Round LDPE Carboys with Spigot Available on GSA/VA Contract for Federal Government customers only.
Collect and dispense distilled water, acids and reagents with Thermo Scientific™ Nalgene™ Round LDPE Carboys with Spigot.
Thermo Scientific™ Nalgene™ Carboy Vent Filter Available on GSA/VA Contract for Federal Government customers only.
Maintain purified water stored in carboys with this PTFE filter, used for venting and solvent filtration applications.
Thermo Scientific™ Nalgene™ Polypropylene Quick Action Spigots Available on GSA/VA Contract for Federal Government customers only.
Incorporate Thermo Scientific™ Nalgene™ Polypropylene Quick-Action Spigots, including an HDPE tubing adapter which accepts tubing with 1/4 or 5/16″ I.D with Carboys.
Thermo Scientific™ Nalgene™ Spigot Closure with Polypropylene Lock Nut and TPE plug Available on GSA/VA Contract for Federal Government customers only.
Replace a spigot when autoclaving liquids in a carboy with the Thermo Scientific™ Nalgene™ Spigot Closure with Polypropylene Lock Nut and TPE Plug to provide a leakproof† seal.
Thermo Scientific™ Nalgene™ LDPE Carboys with Tubulation and Closure Available on GSA/VA Contract for Federal Government customers only.
Perform applications with confidence thanks to the strength, long life and leak resistance of these one-piece molded LPDE carboys with tubulation.
Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers Available on GSA/VA Contract for Federal Government customers only.
Store and transport dry and solid materials, specimens and general lab supplies. These stackable containers feature positive-lock lids that form a tight seal.
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboy with Spigot Available on GSA/VA Contract for Federal Government customers only.
Store and dispense solutions and media using these autoclavable polypropylene carboys, featuring a spigot boss molded into the carboy for convenience and strength.
Thermo Scientific™ Nalgene™ Quick-Action Tefzel™ ETFE Spigots Available on GSA/VA Contract for Federal Government customers only.
Use these replacement spigots for select Thermo Scientific™ Nalgene™ carboys. They are chemically resistant to selected solvents and other chemicals.
Thermo Scientific™ Nalgene™ Round Polycarbonate Clearboy™ with Spigot Available on GSA/VA Contract for Federal Government customers only.
Prepare large volume media and cultures with these round polycarbonate Clearboys. Lighter, safer and more durable than glass. Useful for visual quality inspections.
Qorpak™ LDPE Cubitainers with White PP SturdeeSeal™ PE Foam Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Non-glug construction facilitates flow of contents om continuous uninterrupted streams
Qorpak™ HDPE Quick Serve Tap for Cubitainers Available on GSA/VA Contract for Federal Government customers only.
Used for dispensing contents to other vessels
Bel-Art™ SP Scienceware™ Jerricans Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Built-in spout with distinct air vent for easy decanting and venting.
Bel-Art™ SP Scienceware™ Dispensing Jug Products for Science Education
Dispense directly from 5 gallon graduated jug without lifting or moving.
Bel-Art™ SP Scienceware™ Vacuum Aspirator Bottles Available on GSA/VA Contract for Federal Government customers only.
Offers easy and continuous aseptic vacuum aspiration of liquids, including tissue culture media, supernatants and chemical wastes
Fisherbrand™ Rectangular Carboy Closures
Value your lab can rely on
Bel-Art™ SP Scienceware™ Carboy with Handle and Screw Cap
Thick-walled, high-density polyethylene carboy has easy-grip handle for transporting
Bel-Art™ SP Scienceware™ Jerricans with Spigot Available on GSA/VA Contract for Federal Government customers only.
Space-saving, high-density polyethylene jerricans are a convenient means to dispense without moving
Fisherbrand™ Polypropylene Rectangular Carboys
Value your lab can rely on. Fisherbrand Polypropylene Rectangular Carboys have a space-saving rectangular design is ideal for storing and dispensing lab water and reagents. Built to last. Made from high quality plastic materials. Autoclavable.
Bel-Art™ SP Scienceware™ Autoclavable Polypropylene Carboys Available on GSA/VA Contract for Federal Government customers only.
Optimized for storing and dispensing large batches of culture media, distilled water and other solutions
Bel-Art™ SP Scienceware™ Carboys with Spigot Available on GSA/VA Contract for Federal Government customers only.
Lightweight, durable polyethylene with extra-heavy walls for long life and dependability
Bel-Art™ SP Scienceware™ Jerricans Available on GSA/VA Contract for Federal Government customers only.
Feature a 20cm (8 in.) spout with a built-in air vent for smooth pouring
Qorpak™ HDPE Square Hedpaks Available on GSA/VA Contract for Federal Government customers only.
Integral handle on top face with molded pouring vent and finger recess on bottom of bottle enables easy pouring
Bel-Art™ SP Scienceware™ Dispensing Jug Available on GSA/VA Contract for Federal Government customers only.
Store and dispense common solutions, including janitorial cleaners, deionized / distilled water, or other aqueous liquids. Bel Art™ SP Scienceware™ Dispensing Jug has molded-in gradations.
  spinner