Cellware and Inserts
Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes Available on GSA/VA Contract for Federal Government customers only.
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.
Corning™ BioCoat™ Collagen I Inserts with PET Membrane
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ BioCoat™ Control Insert Available on GSA/VA Contract for Federal Government customers only.
Use alongside BD BioCoat™ inserts when studying effects of the ECM component present on the BD BioCoat cell culture insert
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Tube Formation
Evaluate and quantitate endothelial cell invasion using real-time fluorescence and non-destructive sample preparation
Corning™ BioCoat™ FluoroBlok™ Fibronectin Cell Culture Inserts
Detect migrated cells below membrane using a fluoresence plate reader
EMD Millipore Millicell™ Culture Plate Inserts Available on GSA/VA Contract for Federal Government customers only.
Designed for improved cell differentiation and expanded applications involving analytical cell biology. EMD Millipore Millicell™ Culture Plate Inserts incorporate PTFE Biopore™ membranes with 0.4μm pores. Ready-to-use 30 or 12mm inserts for 6 and 24-well plates.
Corning™ BioCoat™ Matrigel™ Growth Factor Reduced Matrix for Smooth Muscle Cells
Induce differentiation of human and rat aortic smooth muscle cells
Corning™ Transwell™ Multiple Well Plate with Permeable Polyester Membrane Inserts Available on GSA/VA Contract for Federal Government customers only.
Featuring a microscopically transparent polyester membrane that is TC-treated for optimal cell attachment and growth. Corning™ Transwell™ Multiwell Plate with Polyester Membrane Inserts provide better visibility under phase contrast microscopy and allow assessment of cell viability and monolayer formation.
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Invasion
Analyze endothelial cell invasion using fluorescence detection in a simplified and reproducible manner
Corning™ BioCoat™ TC-treated Cultureslide Variety Pack
Choose from different groups of cellware conveniently packaged together in popular combinations
Corning™ BioCoat™ Poly-D-Lysine/Laminin Culture Slide Available on GSA/VA Contract for Federal Government customers only.
Tested for the ability to promote neurite outgrowth with NG108 rat glioma/mouse neuroblastoma cells
Corning™ BioCoat™ Fibronectin Culture Slide, Human
Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
EMD Millipore Millicell™ Culture Plate Inserts: PCF Available on GSA/VA Contract for Federal Government customers only.
Designed for improved cell differentiation and expanded applications involving analytical cell biology
Corning™ BioCoat™ Tumor Invasion Multiwell Plate Available on GSA/VA Contract for Federal Government customers only.
Provides an environment that allows for the assessment of the invasive potential of cells in vitro
Corning™ BioCoat™ Fibronectin, Human Cell Culture Insert
Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
Corning™ BioCoat™ Matrigel™ Invasion Chamber with BD Matrigel Matrix Available on GSA/VA Contract for Federal Government customers only.
Provide rapid and reproducible quantification of cell invasion in vitro. Corning™ BioCoat™ Matrigel™ Invasion Chamber with BD Matrigel Matrix™ cell culture inserts are pre-coated with extracellular matrix proteings for applicatins requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and differentiation.
Corning™ BioCoat™ TC-treated Multiwell Variety Packs
Choose from different groups of cellware conveniently packaged together in popular combinations
Corning™ Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts Available on GSA/VA Contract for Federal Government customers only.
Patented self-centering design provides independent access to both apical and basolateral membranes of the cell monolayer. Corning™ Transwell™Multiple Well Plate with Permeable Polycarbonate Membrane Inserts provide an ideal solution for studying anchorage-dependent and -independent cells.
EMD Millipore Millicell™ Culture Plate Inserts: HATF Available on GSA/VA Contract for Federal Government customers only.
Designed for improved cell differentiation and expanded applications involving analytical cell biology
Corning™ BioCoat™ Fibronectin HTS Multiwell Angiogenesis
Provides a standard format to increase the quality and reproducibility of in vitro endothelial cell migration assays
Corning™ BioCoat™ Intestinal Epithelium Differentiation Environment Kit
Used to create intestinal models that exhibit barrier function for use in transport and absorption studies of drugs, metabolites and infectious diseases of the intestine
Corning™ BioCoat™ Poly-D-Lysine Glass Multiwell Culture Slide Available on GSA/VA Contract for Federal Government customers only.
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ Snapwell™ Culture Inserts Available on GSA/VA Contract for Federal Government customers only.
Designed with a ring supported membrane for use with diffusion or Ussing chambers once cells are grown to confluence. Corning™ Snapwell&trade Culture Inserts are primarily used for transport and electrophysiological studies.
Corning™ BioCoat™ Collagen I Glass Multiwell Culture Slide Available on GSA/VA Contract for Federal Government customers only.
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ BioCoat™ Matrigel™ Invasion Chamber: With GFR Matrigel Matrix Available on GSA/VA Contract for Federal Government customers only.
Provide rapid and reproducible quantification of cell invasion in vitro
  spinner