Flat Sided Cell Culture Flasks and Accessories
Thermo Scientific™ Nunc™ Cell Culture Treated TripleFlasks™ Available on GSA/VA Contract for Federal Government customers only.
Perform large cell mass applications such as drug screening and scale up with these cell-culture treated TripleFlasks.
Wheaton™ CELLine™ Bioreactor Flask
Membrane cell culture flasks ideal for antibody and protein production
Corning™ BioCoat™ Gelatin Rectangular Canted Neck Culture Flasks w/Vented Cap
Pre-coating with gelatin (obtained from porcine gelatin) eliminates time-consuming preparation
Corning™ Costar™ Cell Culture Triangular Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polystyrene for optimum visibility. Corning™ Cell Culture Triangular Flasks provide excellent accessibility to cell surface.
Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap
Pre-coated with Collagen I for improved cell attachment and an increased proliferation rate of transfected mammalian cells, primary cells and stem cells. Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap provides highly consistent and biologically functional precoated surfaces to more closely mimic in vivo environments for cell culture applications.
Corning™ HYPERStack™ Accessories: Bottle Stand Available on GSA/VA Contract for Federal Government customers only.
Accessories for use with Corning HYPERStack products
Corning™ BioCoat™ Poly-D-Lysine Vented Cap Flasks
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ Cell Culture Treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polstyrene for optimum visibility and distortion-free microscopic viewing. Corning™ Cell Culture Treated Flasks are available in a variety of sizes, designs and cap styles to meet laboratory diagnostic needs.
Corning™ HYPERStack™ Accessories: Stack Manipulator Available on GSA/VA Contract for Federal Government customers only.
Accessories for use with Corning HYPERStack products
Kimble™ Kontes™ Nephelo Culture Flasks
For Monitoring in-situ cell/bacterial culture
Corning™ BioCoat™ Fibronectin Rectangular Canted Neck Culture Flask
Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
Corning™ PureCoat™ ECM Mimetic Multi-Flask Available on GSA/VA Contract for Federal Government customers only.
Features multi-layers with vented cap
Corning™ HYPERFlask™ M Cell Culture Vessels Available on GSA/VA Contract for Federal Government customers only.
Designed specifically for manual use, high-yield and high-performance. Specifically designed for manual use. Corning™ HYPERFlask™ M Cell Culture Vessel uses a multi-layer gas permeable growing surface for efficient gas exchange.
Corning™ Cell Culture Untreated Flasks Available on GSA/VA Contract for Federal Government customers only.
Printed with lot numbers for ease in traceability
Corning™ U-Shaped Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Corning™ PureCoat™ Amine Flasks
Positively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays
Corning™ CellBIND™ Surface Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.
Corning™ CELLine Disposable Bioreactor
Cultivation flask for secreted product, recombinant protein expression and high-yield monoclonal antibody (MAb) production
Corning™ PureCoat™ Fibronectin Peptide Cell Culture Flasks with Vented Cap
Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface
Corning™ PureCoat™ Collagen Peptide Cell Culture Flasks with Vented Cap
Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface
Corning™ Cell Culture Flask with Bar Code Available on GSA/VA Contract for Federal Government customers only.
Features nonrepeating bar code for use in automation
Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology. Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask is ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.
Corning™ PureCoat™ Carboxyl Rectangular Canted Neck Cell Culture Flasks with Vented Cap
Negatively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays
Corning™ HYPERStack™ Accessories: Stacking Tray
Accessories for use with Corning HYPERStack products
Corning™ Low Profile Cell Culture Flask Available on GSA/VA Contract for Federal Government customers only.
Unique design reduces stacking height
Corning™ BioCoat™ Laminin Rectangular Canted Neck Cell Culture Flask w/Plug Seal Cap
Tested for their ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells
Corning™ Ultralow Attachment Flasks Available on GSA/VA Contract for Federal Government customers only.
Features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation. Corning™ Ultralow Attachment Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Corning™ U-Shaped Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
  spinner