Plastic Erlenmeyer Flasks
Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure Available on GSA/VA Contract for Federal Government customers only.
Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.
Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
CombineThermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.
Thermo Scientific™ Nalgene™ Polycarbonate Baffled Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Increase mixing efficiency with these polycarbonate flasks, an alternative to glass flasks that produce equivalent growth curves with E. coli and S. cerevisiae.
Thermo Scientific™ Nalgene™ Teflon™ FEP Erlenmeyer Flasks with Closure Available on GSA/VA Contract for Federal Government customers only.
Effectively perform organic solvent and concentrated oxidizing agent work with these flasks, which are unbreakable and resist virtually all chemicals.
Thermo Scientific™ Nalgene™ Polycarbonate Erlenmeyer Flasks with Closure Available on GSA/VA Contract for Federal Government customers only.
Prepare and store culture media using these shatter-proof Nalgene polycarbonate Erlenmeyer flasks -- suitable for culturing techniques. Safer than glass flasks in shakers.
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile Available on GSA/VA Contract for Federal Government customers only.
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.
Thermo Scientific™ Nalgene™ Polycarbonate Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Conduct many culturing techniques and store culture media with Thermo Scientific™ Nalgene™ Polycarbonate Erlenmeyer Flasks.
Corning™ Baffled Polycarbonate Flasks with Caps Available on GSA/VA Contract for Federal Government customers only.
Designed with baffles to provide premium agitation at lower speed for gentler, more efficient aeration. Corning™ Baffled Polycarbonate Flasks with Caps are ideal for shaker culture applications.
PYREX™ Low-Form Culture Flask Available on GSA/VA Contract for Federal Government customers only.
For culturing organisms requiring a large surface area-to-volume ratio
Wheaton™ Trypsinizing Flasks Available on GSA/VA Contract for Federal Government customers only.
Deep fluted sides enhance agitation
PYREX™ Culture Media Flasks Available on GSA/VA Contract for Federal Government customers only.
Suitable for mixing and storing culture media
Kimble™ KIMAX™ Long, Straight-Neck Flask with Capacity Scale
Neck accepts metal or plastic cap or cotton plug
Kimble™ KIMAX™ Baffled Culture Flasks
For fermentation, cell culture growth and cultivation, and genetic engineering procedures
PYREX™ Delong Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Heavy, fire-polished, long necks reduce splashing
Corning™ Disposable Polycarbonate Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
Wheaton™ Polycarbonate Erlenmeyer Shake Flasks
Ideal replacement for glass Erlenmeyer/Shake Flasks
Corning™ Polycarbonate 5L Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Made of noncytotoxic USP Class VI polycarbonate that provides optical clarity and mechanical strength. Corning™ Polycarbonate 5L Erlenmayer Flask are ready-to-use for efficient scale-up of suspension cultures with space efficiency.
PYREX™ Fernbach Flasks Available on GSA/VA Contract for Federal Government customers only.
Fernsbach culture flasks offer wide surface area is ideal for growth of organisms requiring a large surface area-to-volume ratio
Corning™ Two- and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Two-and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
Wheaton™ Nephelo Culture Flask with Cleanout Port and Caps Available on GSA/VA Contract for Federal Government customers only.
Features depressed side arm and side clean-out port
  spinner