Laboratory Plastic Erlenmeyer Flasks
Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
CombineThermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.
Thermo Scientific™ Nalgene™ Polycarbonate Erlenmeyer Flasks with Closure Available on GSA/VA Contract for Federal Government customers only.
Prepare and store culture media using these shatter-proof Nalgene polycarbonate Erlenmeyer flasks -- suitable for culturing techniques. Safer than glass flasks in shakers.
Thermo Scientific™ Nalgene™ Polycarbonate Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Conduct many culturing techniques and store culture media with Thermo Scientific™ Nalgene™ Polycarbonate Erlenmeyer Flasks.
Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure Available on GSA/VA Contract for Federal Government customers only.
Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile Available on GSA/VA Contract for Federal Government customers only.
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.
Thermo Scientific™ Nalgene™ Teflon™ FEP Erlenmeyer Flasks with Closure Available on GSA/VA Contract for Federal Government customers only.
Effectively perform organic solvent and concentrated oxidizing agent work with these flasks, which are unbreakable and resist virtually all chemicals.
Wheaton™ Polycarbonate Erlenmeyer Shake Flasks
Ideal replacement for glass Erlenmeyer/Shake Flasks
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile Available on GSA/VA Contract for Federal Government customers only.
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.
  spinner