Plastic Volumetric Flasks
Corning™ Class B Reusable Polypropylene Volumetric Flasks Products for Science Education
Supplied with tapered polypropylene stopper with an individually calibrated graduation line.
Thermo Scientific™ Nalgene™ Class B Polymethypentene (PMP) Volumetric Flasks with Screw Caps Available on GSA/VA Contract for Federal Government customers only.
Protect against leakage, breakage, and contamination with these volumetric flasks, calibrated “To Contain” and “To Deliver” at 20°C.
Thermo Scientific™ Nalgene™ Class B Polypropylene Copolymer Volumetric Flasks with Closure Available on GSA/VA Contract for Federal Government customers only.
Lighter in weight than glass, these flasks are break-resistant, offer excellent chemical resistance and protect against leakage and contamination.
Corning™ Class B Reusable Polypropylene Volumetric Flasks Available on GSA/VA Contract for Federal Government customers only.
Supplied with tapered polypropylene stopper with an individually calibrated graduation line
Fisherbrand™ PFA Flasks with Screw Caps Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Contain” within Class A tolerances
  spinner