Plastic Squibb Pear Shaped Separatory Funnels
Squibb Pear-Shaped Separatory Funnels Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Good clarity for viewing layers.
United Scientific Supplies Separatory Funnels Products for Science Education
Provide excellent clarity and strength.
Thermo Scientific™ Nalgene™ Teflon™ FEP Separatory Funnels with Closure Available on GSA/VA Contract for Federal Government customers only.
Utilize these chemically inert, translucent and autoclavable funnels which feature a TFE stopcock assembly. Chemical-resistant, leakproof and requires no lubrication.
Thermo Scientific™ Replacement Stopcock Assembly for Nalgene™ Separatory Funnels Available on GSA/VA Contract for Federal Government customers only.
Maintain separatory funnels with this Replacement Stopcock Assembly for Thermo Scientific™ Nalgene™ Separatory Funnels, which ensures a leakproof† fit without lubrication.
Thermo Scientific™ Nalgene™ Polypropylene Separatory Funnels with Closure Available on GSA/VA Contract for Federal Government customers only.
Perform a variety of lab duties with chemically inert, translucent and autoclavable Thermo Scientific™ Nalgene™ Polypropylene Separatory Funnels.
  spinner