Crystallization Microplates
AxyGem™ Crystallography Plate Available on GSA/VA Contract for Federal Government customers only.
With 96 buffer wells and an adjacent protein well
AxyGem Plate is an SBS standard 96-well sitting-drop, high-throughput protein crystallization plate with 96 buffer wells (maximum volume of 250μL) and one adjacent protein well (maximum volume of 4.3μL)
Corning™ CrystalEX 96-Well Protein Crystallization Microplates Available on GSA/VA Contract for Federal Government customers only.
Designed for sitting-drop vapor diffusion high throughput protein crystallization
Corning™ 96-Well Next Generation CrystalEX Protein Crystallization Microplates Available on GSA/VA Contract for Federal Government customers only.
Optimized for high-throughput sitting-drop protein crystallization
Corning™ CrystalEX384-Well Flat Bottom Protein Crystallization Microplate Available on GSA/VA Contract for Federal Government customers only.
The only microplate designed for full automation in crystal screening
  spinner