Non Binding Microplates
Corning™ 96-Well Nonbinding Surface (NBS™) Microplates Available on GSA/VA Contract for Federal Government customers only.
Treated with a Corning proprietary treatment technology to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Corning™ 96-Well Nonbinding Surface (NBS™) Microplates are ideal for reducing proteing and nucleic acid binding at low concentrations, and increasing assay signal to noise.
Corning™ 96-Well Clear Ultra Low Attachment Microplates Available on GSA/VA Contract for Federal Government customers only.
Covalently bonded hydrogel minimizes cell attachment, protein absorption, enzyme activation and cellular activation. Corning™ 96-Well Clear Ultra Low Attachment Microplates allow for lentivirus transduction of human osteoclast precursor cells.
Corning™ Costar™ Clear Multiple Well Plates Available on GSA/VA Contract for Federal Government customers only.
Untreated surface is hydrophobic in nature and binds bio-olecules through passive interactions. Corning™ Costar™ Clear Multiple Well Plates provide an ideal solution for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.
  spinner