UV Microplates
Corning™ UV-Transparent Microplates Available on GSA/VA Contract for Federal Government customers only.
Designed with a UV-transparent well bottom for determining protein and nucleic acid concentrations. Corning™ UV-Transparent Microplates allows UV absorbance readings with low background, especially at 260 to 280nm.
  spinner