Milk Bacteriological Pipets
Kimble™ KIMAX™ Calibrated “To Deliver” Babcock Bottle Test Pipet
Milk test; Calibrated to deliver for no blow-out
Kimble™ Sterile Disposable Polystyrene Milk Bacteriological Pipets
1.1mL polystyrene pipet with plugged mouth end
Kimble™ KIMAX™ Brand Reusable Milk Test Pipet for Gerber Bottles
Babcock pipet with special rubber washer; brown stain scale.
Kimble™ KIMAX™ Reusable Cream Pipets
Opening at tip larger than ordinary milk pipets.
  spinner