Immunology Tubes
Corning™ Cluster Tube System Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.
Thermo Scientific™ Immuno Tubes and Stoppers Available on GSA/VA Contract for Federal Government customers only.
Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.
  spinner