Glass Cell Culture Tubes
Kimble™ Rim Top Mark-M™ Borosilicate Glass Culture Tubes with Marking Labels Available on GSA/VA Contract for Federal Government customers only.
Permanent ceramic label, fused to the glass located 3/4 in. from the open end
Fisherbrand™ Reusable Glass Rim Top Culture Tubes Available on GSA/VA Contract for Federal Government customers only.
Uniform wall thickness for maximum heat transfer and chemical resistance
PYREX™ VISTA™ Rimless Culture Tubes Products for Science Education
Offer convenient plugging and rack storage.
Fisherbrand™ Disposable Borosilicate Glass Tubes with Plain End Available on GSA/VA Contract for Federal Government customers only.
Use these tubes for tissue culture and clinical chemistry applications. Fisherbrand™ Disposable Borosilicate Glass Tubes with Plain End are made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda lime glass.
PYREX™ Disposable Rimless Flat-Bottom Glass Tubes Available on GSA/VA Contract for Federal Government customers only.
Ideal for plant tissue culture and general culture work
PYREX™ VISTA™ Rimless Reusable Glass Culture Tubes Available on GSA/VA Contract for Federal Government customers only.
Functionality, economy, and quality
PYREX™ Disposable Round-Bottom Rimless Glass Tubes Available on GSA/VA Contract for Federal Government customers only.
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
PYREX™ Culture Tubes with Threaded End and Marking Spot Available on GSA/VA Contract for Federal Government customers only.
For tissue culture and general bacteriological work
Pyrex™ Round Bottom Threaded Culture Tubes Available on GSA/VA Contract for Federal Government customers only.
Made from borosilicate glass for use with samples sensitive to leaching from plastic tubes. Corning™ Pyrex™ Round Bottom Threaded Culture Tubes are designed for tissue culture and general bacteriological work.
Kimble™ ValueWare™ Culture Tubes With Marking Spots Products for Science Education
With fire-polished top rim and uniform wall for maximum heat transfer and chemical resistance.
Kimble™ ValueWare™ Culture Tubes with Rubber Lined Phenolic Caps Products for Science Education
Mineral-filled cap to withstand repeated autoclaving.
Kimble™ KIMAX™ Culture Tube Screw Cap and Rubber Liner Products for Science Education
Convenient spot on tube for marking identification.
PYREX™ VISTA™ Reusable Glass Tubes with Phenolic Screw Caps Available on GSA/VA Contract for Federal Government customers only.
Supplied with deep-form phenolic caps to facilitate handling and sealing after autoclaving
Kimble™ Kontes™ Screw Thread Culture Tubes with Unattached Rubber Lined Caps
Uniform wall thickness for maximum heat transfer and heat resistance
Kimble™ Disposable Glass Culture Tube
Round bottomed with threaded neck
  spinner