Plastic Scintillation Vials
Wheaton™ 20mL PET Liquid Scintillation Vials, With Separate Caps Available on GSA/VA Contract for Federal Government customers only.
Offer low permeability to solvents and minimal background counts
Wheaton™ Liquid Scintillation Vials Available on GSA/VA Contract for Federal Government customers only.
Maintain the integrity of seals with a variety of cap liners. Wheaton™ Liquid Scintillation Vials are manufactured from rigid yet flexible HDPE plastic.
Fisherbrand™ 7mL HDPE Scintillation Vials Available on GSA/VA Contract for Federal Government customers only.
Smaller capacity vial saves in solvent use and disposal
Wheaton™ Glass 20mL Scintillation Vials: Polypropylene Caps
Maintain integrity of seal with variety of cap liners. Wheaton Science Products™ Glass 20mL Scintillation Vials: Polypropylene Caps are Chemically inert, suitable for strong solvents and high-purity chemicals.
Kimble™ 20mL Glass Screw-Thread Scintillation Vials
Manufactured from clear KG-33 borosilicate glass
Fisherbrand™ 4, 6, and 8mL HDPE ScintiVials™ Available on GSA/VA Contract for Federal Government customers only.
High-density polyethylene (HDPE) vials with white polypropylene caps
Wheaton™ Liquid Scintillation Vials Available on GSA/VA Contract for Federal Government customers only.
Provides an economical alternative to standard scintillation vials. Wheaton™ Liquid Scintillation Vials are manufactured from rigid yet flexible HDPE plastic.
Fisherbrand™ Borosilicate Glass Scintillation Vials, with white polypropylene caps, 20mL
Use these general purpose vials with all common counters. Fisherbrand™ 20mL Borosilicate Glass Scintillation Vials with white polypropylene caps feature potassium-free glass composition which provides consistently low activity counts and excellent light transmission.
Kimble™ 7mL Solvent-Saver™ Scintillation Vials
Use same volume of radioactive sample as 20mL vials for savings in solvent and solvent disposal costs
Kimble™ Polyethylene Scintillation Vials Available on GSA/VA Contract for Federal Government customers only.
Especially suitable for low activity counting
Wheaton™ Glass 20mL Scintillation Vials and Urea Caps
Maintains integrity of seal with variety of cap liners. Wheaton Science Products™ Glass 20mL Scintillation Vials and Urea Caps are suitable for strong solvents and high-purity chemicals.
Wheaton™ HDPE Sampule™ Available on GSA/VA Contract for Federal Government customers only.
Economical alternative to standard scintillation vials
Fisherbrand™ 7mL Borosilicate Glass Scintillation Vial
Smaller capacity vial saves in solvent use and disposal
Fisherbrand™ Scintillation Vials Available on GSA/VA Contract for Federal Government customers only.
High-density polyethylene vials with threaded cap
Wheaton™ 20mL PET Liquid Scintillation Vials, With Caps Attached Available on GSA/VA Contract for Federal Government customers only.
For optimal performance in the laboratory.  Wheaton™ 20mL PET Liquid Scintillation Vials, With Caps Attached offer low permeability to solvents and minimal background counts.
Simport™ Scientific 6.5mL HDPE Snaptwist™ Scintillation Vial
With polypropylene cap.Simport™Scientific 6.5mL HDPE Snaptwist™ Scintillation Vials are designed for use with liquid scintillation and gamma counting, chromatography, sample storage and culturing.
  spinner