Tissue Grinders
PYREX™ PTFE Pestle Tissue Grinder with Steel Shaft Available on GSA/VA Contract for Federal Government customers only.
Recommended for soft tissues found in the liver and brain
Kimble™ Kontes™ Potter-Elvehjem Tissue Grinder
Large volume with a finely machined PTFE pestle
Kimble™ Kontes™ Pellet Pestle™ Available on GSA/VA Contract for Federal Government customers only.
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer Available on GSA/VA Contract for Federal Government customers only.
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
Wheaton™ Tapered Tissue Grinders With PTFE Pestles Available on GSA/VA Contract for Federal Government customers only.
Tapered tissue grinder with a PTFE pestle for soft tissues
Kimble™ Kontes™ Duall™ Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Combined conical and cylindrical surfaces improve grinding efficiency
Wheaton™ Potter-Elvehjem Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Choose grinders with or without plastic safety coating
Kimble™ Kontes™ Closed System Sterile Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Minimizes the risk of personal contact with samples
Fisherbrand™ Disposable Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Closed tissue grinder system minimizes sample contact by aerosols and sample overflow
Wheaton™ Grinding Chamber Replacement Parts For Tapered Tissue Grinders With PTFE Pestles Available on GSA/VA Contract for Federal Government customers only.
Tapered tissue grinders replacement grinding chamber for fluoropolymer resin pestle
Wheaton™ Micro Tissue Grinders
Used in extremely precise work when delicate hand operation is required
PYREX™ Potter-Elvehjem Tissue Grinder with PTFE Pestle Available on GSA/VA Contract for Federal Government customers only.
Used in preparing small samples for pathological and biochemical studies
Wheaton™ Tapered Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Improved design with longer tapered surface on both the mortar and pestle
PYREX™ Glass Pestle Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Consist of glass tube and pestle with clearance of 0.15mm
Wheaton™ Dounce Dura-Grind™ Tissue Grinder Available on GSA/VA Contract for Federal Government customers only.
An alternative to glass tissue grinders
Wheaton™ Tenbroeck Tissue Grinder
With expanded reservoir and pouring lip
Wheaton™ Dounce Tissue Grinders
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
Kimble™ Kontes™ Dounce Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Designed for cellular work in which the nucleus remains intact after homogenization
Kimble™ Kontes™ All-Glass Potter-Elvehjem Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Designed for motor drive using 788000 Stirrer Adapter
Fisherbrand™ Pellet Pestles™ Available on GSA/VA Contract for Federal Government customers only.
Resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
  spinner