Electrophoresis Consumables and Reagents
Thermo Scientific™ NoLimits DNA Fragment 700 bp Available on GSA/VA Contract for Federal Government customers only.
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
Thermo Scientific™ NoLimits DNA Fragment 850 bp Available on GSA/VA Contract for Federal Government customers only.
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
Thermo Scientific™ GeneRuler 100 bp DNA Ladder, Ready-to-Use 100 to 1000 bp Available on GSA/VA Contract for Federal Government customers only.
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.
Acrylamide / N,N'-Methylenebisacrylamide 19:1, for biochemistry, 40% mix solution in, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acrylamide / N, N'-Methylenebisacrylamide 19:1 CAS: 110-26-9 CAS: 7732-18-5 CAS Min %: 59.0 CAS Max %: 61.0 CAS: 110-26-9 CAS: 79-06-1 Purity Grade: For biochemistry MDL Number: MFCD00080848
Thermo Scientific™ NoLimits 1000 bp DNA Fragment Available on GSA/VA Contract for Federal Government customers only.
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
Agarose 0.8% Products for Science Education
Chemical Name or Material: Agarose CAS: 9012-36-6 Synonym: Powder
Thermo Scientific™ pUC19 DNA/MspI (HpaII) Marker 23 Available on GSA/VA Contract for Federal Government customers only.
Perform sizing and approximate quantification of small linear double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels.
Thermo Scientific™ GeneRuler DNA Ladder Mix, ready-to-use Available on GSA/VA Contract for Federal Government customers only.
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels.
Thermo Scientific™ GeneRuler™ Low Range DNA Ladder Available on GSA/VA Contract for Federal Government customers only.
Perform fast sizing and approximate quantification of low molecular weight double-stranded DNA on agarose gels.
Thermo Scientific™ NoLimits DNA Fragment 1200 bp Available on GSA/VA Contract for Federal Government customers only.
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
Thermo Scientific Pierce™ Power Staining Kits
Get fast Coomassie staining of protein gels with Thermo Scientific™ Pierce™ Power Staining Kits, optimized to work exclusively with the Pierce Power Stain Cassette and Pierce Power Station.
Agarose 2% Products for Science Education
Chemical Name or Material: Agarose CAS: 9012-36-6 Synonym: Powder
Thermo Scientific™ O'RangeRuler 200 bp DNA Ladder, Ready-to-Use 200-3000 bp Available on GSA/VA Contract for Federal Government customers only.
Perform precise sizing of PCR products and other double-stranded DNA fragments in agarose or non-denaturing polyacrylamide gels with this DNA step ladder of 200 bp increments.
Thermo Scientific™ NoLimits 100 bp DNA Fragment Available on GSA/VA Contract for Federal Government customers only.
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
Thermo Scientific™ RiboRuler Low Range RNA Ladder 100-1000 bp Available on GSA/VA Contract for Federal Government customers only.
Size and quantify RNA with Thermo Scientific™ RiboRuler Low Range RNA Ladder, a mixture of 7 chromatography purified single-stranded RNA transcripts (100 to 1000 bp long).
N,N'-Methylenebisacrylamide, ≥99%, for biochemistry, for electrophoresis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N';-Methylenebisacrylamide Name Note: For electrophoresis, For biochemistry CAS: 110-26-9 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For biochemistry Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C7H10N2O2 Linear Formula: (H2C=CHCONH)2CH2 Formula Weight: 154.17 MDL Number: MFCD00008625 Synonym: MBA
Thermo Scientific™ GeneRuler™ 50bp DNA Ladder Available on GSA/VA Contract for Federal Government customers only.
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels with this mixture of chromatography-purified individual DNA fragments.
Silica gel, C18-RP, 17%C, ca.0,8mmol/g, part. size: 40-63μ, ACROS Organics™
Chemical Name or Material: Silica gel, C18-RP, 17%C CAS: 71889-02-6 Synonym: Octadecylsilicate
Thermo Scientific™ NoLimits DNA Fragment 1500 bp Available on GSA/VA Contract for Federal Government customers only.
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
Agarose LE, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Agarose medium-low EEO CAS: 9012-36-6 Purity Grade: For electrophoresis MDL Number: MFCD00081294
Thermo Scientific™ Spectra™ Multicolor Broad Range Protein Ladder
A 4-color protein standard containing 10 prestained proteins (10 to 260kDa) for use as size standards in gel electrophoresis and Western blotting.
Thermo Scientific™ NoLimits DNA Fragment 17,000 bp Available on GSA/VA Contract for Federal Government customers only.
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
Thermo Scientific™ O'RangeRuler 500 bp DNA Ladder, Ready-to-Use 500-6000 bp Available on GSA/VA Contract for Federal Government customers only.
Perform precise sizing of PCR products and other double-stranded DNA fragments in agarose or non-denaturing polyacrylamide gels with this DNA step ladder of 500 bp increments.
Acrylamide, 98.5%, extra pure, ACROS Organics™
Chemical Name or Material: Acrylamide CAS: 79-06-1 Purity Grade: Extra Pure Assay: ≥98.0% (GC) Assay Percent Range: 98.5% Molecular Formula: C3H5NO Linear Formula: H2C=CHCONH2 Formula Weight: 71.08 Synonym: 2-Propenamide
Thermo Scientific™ NoLimits DNA Fragment 250 bp Available on GSA/VA Contract for Federal Government customers only.
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
  spinner