Dye Based Protein Gel Stains
Edvotek™ Protein InstaStain™ sheets Products for Science Education
Stain gels faster than conventional methods avoiding large amounts of liquid stain and waste disposal.
Edvotek™ InstaStain™ Blue Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Environmentally friendly, avoiding large amounts of liquid stain and waste disposal.
Thermo Scientific™ GelCode™ Blue Stain Reagent
Achieve nanogram-level detection and results that exceed those of typical coomassie stains, all without a destaining step.
Thermo Scientific™ GelCode™ Blue Safe Protein Stain
Rapidly detect proteins in polyacrylamide gels with this one-step, ultrasensitive stain formulated to avoid hazardous shipping costs.
Thermo Scientific™ Pierce™ Coomassie Brilliant Blue R-250 and G-250 Dyes
Prepare colorimetric protein gel stains using purified powdered preparations of the two most common forms of coomassie dye as the base.
Thermo Scientific™ Imperial™ Protein Stain
Detect proteins in the low-nanogram range with this ready-to-use, in-gel, colorimetric stain formulated to deliver fast and sensitive SDS-PAGE staining.
Thermo Scientific™ Pierce™ Glycoprotein Staining Kit
Rapidly stain glycosylated proteins in polyacrylamide gels using the periodic acid-Schiff (PAS) method in this convenient kit.
Thermo Scientific™ Midi Gel Incubation Trays
Incubate midi-sized protein gels and Western blot membranes for staining, washing, probing and other steps of detection procedures using these convenient plastic Thermo Scientific™ Midi Gel Incubation Trays.
Thermo Scientific™ Mini Gel Incubation Trays
Incubate mini-sized protein gels and Western blot membranes for staining, washing, probing and other steps of detection procedures using these convenient plastic Thermo Scientific™ Mini Gel Incubation Trays.
Thermo Scientific™ Pierce™ Reversible Protein Stain Kits for Membranes
Reversibly stain proteins on nitrocellulose or PVDF membrane after transfer from polyacrylamide gels with this sensitive Ponceau S alternative.
Thermo Scientific™ Pierce™ PageBlue™ Protein Staining Solution
A sensitive formulation of colloidal coomassie G-250 dye for endpoint staining of proteins in polyacrylamide gels and on PVDF membranes without methanol or acetic acid.
Thermo Scientific Pierce™ Power Staining Kits
Get fast Coomassie staining of protein gels with Thermo Scientific™ Pierce™ Power Staining Kits, optimized to work exclusively with the Pierce Power Stain Cassette and Pierce Power Station.
Thermo Scientific™ Pierce™ Zinc Reversible Protein Stain Kit
Visualize proteins in polyacrylamide gels immediately after electrophoresis and then erase the stain for subsequent transfer or band extraction.
Fisher BioReagents™ EZ-Run™ Protein Gel Staining Solution
Ready-to-use solution for highly sensitive staining of proteins separated in polyacrylamide gels
Molecular Probes™ CandyCane™ Glycoprotein Molecular Weight Standards, Unstained Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
CandyCane glycoprotein molecular weight standards separate into alternating bands of glycosylated and nonglycosylated proteins during polyacrylamide gel electrophoresis
Molecular Probes™ PRO-Q™ Diamond Phosphoprotein Gel Stain Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Eliminates the need for blotting or phosphoprotein specific antibodies and Western blot analysis
Molecular Probes™ SYPRO™ Tangerine Protein Gel Stain (5000X Concentrate in DMSO) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Less hazardous, less waste and stained proteins can be viewed with a standard UV or blue-light transilluminator or with a laser scanner
Invitrogen™ Novex™ Colloidal Blue Staining Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Five times more sensitivity than Coomassie™ R-250— and can detect as little as 10ng of BSA
Molecular Probes™ SYPRO™ Protein Gel Stain Starter Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Makes it easy for first-time users to try three SDS-PAGE stains
Invitrogen™ Novex™ InVision™ His-Tag In-Gel Stain Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
One ready-to-use solution that does not require mixing, dilution, or solution preparation
Molecular Probes™ WesternDot™ 625 Goat Anti-Mouse Western Blot Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Detection performance and ease-of-use exceed existing ECL-based methods
Molecular Probes™ PRO-Q™ Diamond Phosphoprotein Gel Destaining Solution Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Eliminates the need for blotting or phosphoprotein specific antibodies and Western blot analysis
Invitrogen™ Novex™ InVision™ His-Tag In-Gel Staining Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
One ready-to-use solution that does not require mixing, dilution, or solution preparation
Molecular Probes™ WesternDot™ 625 Goat Anti-Rabbit Western Blot Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Detection performance and ease-of-use exceed existing ECL-based methods
Invitrogen™ Novex™ SYPRO™ Ruby Protein Blot Stain Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Compatible with immunostaining (Western blotting), microsequencing and mass spectrometry
Invitrogen™ Novex™ SimplyBlue™SafeStain Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Less hazardous and does not require methanol or acetic acid fixatives or destains
Molecular Probes™ SYPRO™ Orange Protein Gel Stain (5,000X Concentrate in DMSO) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Fast, one-step staining protocol requiring no destaining
  spinner