Gel Casting and Polymerization Reagents
Thermo Scientific™ Pierce™ Ammonium Persulfate (APS)
Catalyze acrylamide and bisacrylamide polymerization when preparing polyacrylamide gels for electrophoresis with APS oxidizing agent.
Thermo Scientific™ Pierce™ Tetramethylethylenediamine (TEMED)
Efficiently catalyze polyacrylamide gel polymerization in SDS-PAGE using TEMED.
R&D Systems Agarose Cleaning and Storage Kit
Contains the optimal formulations for cleaning and storage of agarose affinity matrices
Lonza GelBond™ PAG Film for Acrylamide Gels
Transparent, flexible polyester film supports polyacrylamide or MDE™ gels through electrophoresis, fixing, staining, destaining, and drying procedures
  spinner