Nucleic Acid Fluorescent Gel Stains
Edvotek™ Electrophoresis Reagent Packages Products for Science Education
Edvotek package contains enough gel loading solution is provided to prepare 200 samples
Edvotek™ Reagents for Electrophoresis: FlashBlue Plus DNA Staining System Products for Science Education
Provides optimized solution to shorten time for staining and destaining steps.
Edvotek™ Electrophoresis Reagent Package with Methylene Blue Plus Products for Science Education
This Electrophoresis Reagent package contains enough reagents to perform up to 40 electrophoresis experiments with the EDVOTEK Electrophoresis Apparatus Model #M6, M12 or M36 - See more at: http://www.edvotek.com/604#sthash.yyLPiNKb.dpuf
Invitrogen™ SYBR™ Gold Nucleic Acid Gel Stain (10,000X Concentrate in DMSO) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Extremely sensitive fluorescent stain for detection of nucleic acids
Invitrogen™ SYBR™ Safe™ DNA Gel Stain Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Highly sensitive stain for visualization of DNA in agarose or acrylamide gels
SlowFade™ Gold Antifade Mountant Non-distribution product as customer accommodation.
Liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides
Invitrogen™ SYBR™ Green I Nucleic Acid Gel Stain - 10,000X concentrate in DMSO Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Extremely sensitive stain for detecting double-stranded DNA (dsDNA) in agarose and polyacrylamide gels
Invitrogen™ SYBR™ Safe™ DNA Gel Stain in 0.5X TBE Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Developed specifically for reduced mutagenicity – to be safer than ethidium bromide for staining DNA in agarose or acrylamide gels
Invitrogen™ SYBR™ Safe™ DNA Gel Stain Starter Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Developed specifically for reduced mutagenicity – as a safer alternative to ethidium bromide for staining DNA in agarose or acrylamide gels
Invitrogen™ SYBR™ Green Nucleic Acid Gel Stain Starter Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Everything needed for first time users to use SYBR gel stains
Invitrogen™ UltraPure™ Ethidium Bromide, 0.625mg/mL Non-distribution product as customer accommodation.
Fluorescent dye used to detect nucleic acids in agarose gels and cesium chloride (CsCl) gradient
EMD Millipore Calbiochem™ FASTsilver™ Gel Staining Kit Available on GSA/VA Contract for Federal Government customers only.
Rapid and sensitive method for detecting protein and nucleic acids fractionated by PAGE
EMD Millipore Eriochrome Black T
Meets ACS specifications
Promega™ Molecular Grade Ethidium Bromide Solution
Molecular grade, fluorescent dye suitable for staining nucleic acids after electrophoresis or in cesium chloride gradients
Innovating Science™ DNA Stain (20X)
Stain and Destain in 40 Minutes. This highly sensitive stain will stain a gel in about 20 minutes and destain in 20 minutes.
Lonza SYBR™ Green I Nucleic Acid Gel Stain
For the detection of dsDNA and oligonucleotides in agarose and polyacrylamide gels
Lonza SYBR™ Green II Nucleic Acid Gel Stain
For the detection of ssDNA and RNA in agarose and polyacrylamide gels
  spinner