Protein Molecular Weight Markers
Thermo Scientific™ PageRuler™ Unstained Low Range Protein Ladder
A mixture of seven purified proteins and peptides (3.4 to 100kDa) for use as size standards in peptide and small protein gel electrophoresis.
Thermo Scientific™ Spectra™ Multicolor Broad Range Protein Ladder
A 4-color protein standard containing 10 prestained proteins (10 to 260kDa) for use as size standards in gel electrophoresis and Western blotting.
Thermo Scientific™ PageRuler™ Plus Prestained 10-250kDa Protein Ladder
Mixture of 9 blue-, orange- and green-stained proteins (10 to 250kDa) used as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
Thermo Scientific™ PageRuler™ Prestained NIR Protein Ladder
A mixture of 10 proteins (11 to 250KDa) that are blue-stained and fluor-labeled for near-IR fluorescent visualization and protein sizing following electrophoresis.
Thermo Scientific™ SuperSignal™ Molecular Weight Protein Ladders
Ready-to-use mix of recombinant proteins (20 to 150K) with IgG-binding sites for chemiluminescent, fluorescent, chromogenic or other detection methods.
Thermo Scientific™ Pierce™ Unstained Protein Molecular Weight Marker
A mixture of seven native proteins (14.4kDa to 116kDa) for use as size standards in protein electrophoresis (SDS-PAGE).
Thermo Scientific™ PageRuler™ Unstained Broad Range Protein Ladder
A mixture of 11 proteins (5 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
Thermo Scientific™ PageRuler™ Unstained High Range Protein Ladder
A mixture of 8 unstained proteins (60 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
Thermo Scientific™ PageRuler™ Prestained 10-180kDa Protein Ladder
Mixture of 10 blue-, orange- and green-stained proteins (10 to 180kDa) used as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
Thermo Scientific™ Spectra™ Multicolor Low Range Protein Ladder
A mixture of six proteins and synthetic peptides (1.7 to 40kDa) for use as size standards for peptides and small proteins in gel electrophoresis and Western blotting.
Thermo Scientific™ Spectra™ Multicolor High Range Protein Ladder
A mixture of eight blue-, green- and orange-stained proteins (40 to 300kDa) for use as size standards for high MW proteins in gel electrophoresis and Western blotting.
Thermo Scientific™ PageRuler™ Unstained Protein Ladder
A mixture of seven native proteins (14.4 to 116kDa) for use as size standards in protein electrophoresis (SDS-PAGE).
Thermo Scientific™ Pierce™ Prestained Protein Molecular Weight Marker
A mixture of six blue-stained natural proteins (20 to 120kDa) for use as size standards in protein electrophoresis (SDS-PAGE).
Fisher BioReagents™ EZ-Run™ Rec Protein Ladder Available on GSA/VA Contract for Federal Government customers only.
Mixture of 14 highly purified recombinant proteins optimized for precise sizing of proteins by SDS-PAGE and Western blotting
Fisher BioReagents™ EZ-Run™ Protein Marker Available on GSA/VA Contract for Federal Government customers only.
Mixture of seven purified proteins optimized to produce uniform, compact bands when analyzed by SDS-PAGE and stained with Coomassie™ Blue
Fisher BioReagents™ EZ-Run™ Prestained Rec Protein Ladder Available on GSA/VA Contract for Federal Government customers only.
Comprised of 10 recombinant proteins covalently coupled to a blue chromophore plus 10 and 72kDa reference bands tagged with green and orange dyes, respectively
Fisher BioReagents™ EZ-Run™ Prestained Protein Marker Available on GSA/VA Contract for Federal Government customers only.
Ideal for monitoring protein separation during electrophoresis without staining and for verifying electroblotting of proteins onto membranes
Invitrogen™ Novex™ Sharp Unstained Protein Standard Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed for accurate, easy and convenient molecular weight estimation of a wide range of molecular weight proteins during SDS-PAGE and western blotting
Invitrogen™ Novex™ SeeBlue™ Plus2 Pre-stained Protein Standard Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Allows you to easily visualize protein molecular weight ranges during electrophoresis and quickly evaluate Western transfer efficiency
Invitrogen™ Novex™ E-PAGE™ MagicMark™ Unstained Protein Standard Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed for use with pre-cast E-PAGE™ Electrophoresis System
Invitrogen™ Novex™ HiMark™ Pre-stained Protein Standard Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed for analysis of high molecular weight proteins on NuPAGE™ Novex™ Tris-Acetate gels
Novex™ BenchMark™ Protein Ladder, Unstained Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Molecular weight standard for SDS-PAGE
Invitrogen™ Novex™ SeeBlue™ Pre-stained Protein Standard Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Allows you to easily monitor the progress of your gel run and is an excellent tool for judging Western transfer efficiency
Invitrogen™ Novex™ MagicMark™ XP Western Protein Standard, Unstained Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Specifically designed for easy and convenient protein molecular weight estimation directly on Western blots
Invitrogen™ Novex™ NuPAGE™ Large Protein Sensitive Staining Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Specifically designed for large protein analysis on NuPAGE Novex Tris-Acetate Gels under denaturing conditions
Invitrogen™ Novex™ BenchMark™ Fluorescent Protein Standard Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Consists of fluorescent dye-conjugated proteins, ranging in size from approx. 11-155kDa
Invitrogen™ Novex™ Sharp Pre-stained Protein Standard Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed for accurate, easy, and convenient molecular weight estimation of a wide range of molecular weight proteins during SDS-PAGE and Western blotting
Invitrogen™ Novex™ NativeMARK™ Unstained Protein Standard, Unstained Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Ready-to-use protein standard designed for molecular weight estimation of proteins using native gel electrophoresis
Invitrogen™ Novex™ E-PAGE™ SeeBlue™ Pre-stained Standard Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed for use with pre-cast E-PAGE™ Electrophoresis System
  spinner