IEF Electrophoresis Systems
ZOOM™ IPGRunner™ Mini-Cell Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Oil-free, trouble-free 2D electrophoresis in a minigel format
GE Healthcare Immobiline™ DryStrip Kit and Components Available on GSA/VA Contract for Federal Government customers only.
Precast Immobiline DryStrip gels for running the first dimension of a 2-D electrophoresis experiment with the Multiphor II electrophoresis system
GE Healthcare Immobiline™ DryPlate for IEF Available on GSA/VA Contract for Federal Government customers only.
Obtain reproducible IEF separations with dry, precast gels containing immobilized pH gradients
GE™ Ettan IPGphor 3 Isoelectric Focusing Unit
Fully-integrated system for high throughput, high protein-loading capacity.
Hoefer™ IEF100 IEF First-Dimension Unit Available on GSA/VA Contract for Federal Government customers only.
Versatile unit accepts up to six 24cm IPG strips or twelve 7cm IGP strips using the dual electrode assembly included
  spinner