Photodocumentation Systems
Edvotek™ White Light Box Products for Science Education
Safely enhance the visualization of DNA stained with FlashBlue™, proteins stained with Coomassie Blue and autoradiograms with the Edvotek White Light Box.
Edvotek™ Personal Digital Photodocumentation Systems for UV or White Light
Combination of easy-to-use digital camera and specially designed hood provides low-cost alternative for gel photos.
UVP PhotoDoc-It™ Imaging System Accessories
Paper/Ink Kit for this high resolution, digital camera for easy gel documentation
UVP ChemiDoc-It™2 510 Imagers
For extensive quantitative analysis of chemiluminescent blots and fluorescent gels
Fotodyne™ FOTO/Analyst Workstation and System Filters Products for Science Education
Special narrow bandpass interference filters are required for optimal image acquisition because CCD cameras are more sensitive to ultraviolet and infrared light than traditional Polaroid cameras
UVP MultiDoc-It™ Imaging Systems
Combines digital color camera, darkroom enclosure and transilluminator for digital imaging of gels, plates, and membranes in multiple-user laboratories
Spectronics Corporation Ultraviolet Viewing Cabinets
Designed for viewing electrophoresis gels and TLC plates
Fotodyne Thermal Printer for Fotodyne Digital Imaging Systems
SONY Digital Grayscale Printer for use with the FOTO/Analyst Express systems
Spectronics Corporation Ultraviolet Viewing Cabinets: Darkroom
Designed for viewing electrophoresis gels and TLC plates
UVP DigiDoc-It™ Imaging Systems
Economical gel imaging systems with digital color camera, compact hood, high-performance transilluminator, capture software and sample viewer, captures high quality gel images
Aushon CiraScan™ Imaging and Analysis System
Complete system includes an improved CCD imager with custom optics designed specifically for quantitative chemiluminescent protein arrays
UVP BioSpectrum 810 Imaging System
Advanced, automated image capture of gels and blots for multiple applications
Fotodyne FOTO/Analyst Express Zoom Lens Workstation
Continuous zoom lens gives you much more flexibility than typical digital cameras
UVP iBox™ Explorer™2 Imaging Microscope
Macroscopic to microscopic scale invivo imaging with deeper interrogation of fluorescent signal in real time
Aushon Cirasoft™ Analyst Software
Offers the flexibility to analyze both small and large protein arrays
UVP iBox™ Scientia™ In Vivo 610 Imaging Systems
Non invasive system detects fluorescent probes in visible to near infrared (NIR) to ultra violet wavelength ranges for in vivo small animal imaging
UVP Large Chromato-Vue™ Viewing Cabinets
Cabinets designed for long life and rugged use in a wide range of applications
UVP PhotoDoc-It™ Imaging Systems
Economical and compact photodocumentation systems capture color images for basic gel documentation
UVP GelDoc-It2 310 Imaging System
Captures high resolution images and facilitates quantitative analysis of fluorescent gels
UVP PhotoDoc-It 60 Imaging Systems Products for Science Education
Capture, print and save color images for basic gel documentation with this compact, economical laboratory package.
UVP Large Chromato-Vue™ Viewing Cabinet Curtains
Curtains for large Chromato-Vue viewing cabinets
Fotodyne™ FOTO/Analyst™ Apprentice System: Transilluminators Available on GSA/VA Contract for Federal Government customers only.
Transilluminators for use with Fotodyne FOTO/Analyst Apprentice System
UVP Colony Doc-It™ Imaging Station
Visualize white light and fluorescent colonies
UVP GelMax Imager
View, capture, analyze and document precast and mini-gel data
UVP ChromaDoc-It™ TLC Imaging System
System with 5.1 megapixel digital color camera, viewing cabinet with overhead UV for TLC color documentation and imaging
UVP Mini Chromato-Vue™ Viewing Cabinet Curtains
Cabinet with contrast control filter that absorbs UV energy and provides a compact darkroom environment for viewing materials
UVP PhotoDoc-It™ Imaging System
UVP PhotoDoc-It™ Imaging System capture color images for basic gel documentation.
UVP Mini Chromato-Vue™ Viewing Cabinets
Provides a compact darkroom environment for viewing materials. UVP™ Mini Chromato-Vue™ Viewing Cabinets include a contrast control filter that absorbs UV energy to protect the eyes.
  spinner