Transilluminators
FisherBiotech™ Transilluminator Safety Cover Products for Science Education
Safety cover for FisherBiotech Transilluminator
Edvotek™ Midrange UV Transilluminator Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Midrange (302nm) UV transilluminator designed to visualize DNA stained with ethidium bromide.
Dynamic Diagnostics Wee Sight™ Transilluminator
Helps to accurately locate the tiny veins in a neonate's small limbs
UVP Benchtop UV Transilluminators: Single UV
New design features upgraded components for increased efficiency, performance and functionality
UVP Benchtop UV Transilluminators: 2UV
New design features upgraded components for increased efficiency, performance and functionality
UVP High-Performance UV Transilluminators
Exclusive 25-watt tubes provide high intensity performance and uniform UV illumination plus sensitivity on stained gels
UVP Benchtop UV Transilluminators: Single UV, Variable Intensity
New design features upgraded components for increased efficiency, performance and functionality
UVP UV Light Converter Plates
Plate is specially designed to easily fit on the transilluminator filter surface
UVP Filter Assemblies for White Light Transilluminators
Replacement filters for TW-26 and TW-43 models white light transilluminators
UVP UV/White Light 8-Watt Transilluminator Products for Science Education
UVP UV/White Light 8-Watt Transilluminator is newly-designed for better performance and efficiency.
Fotodyne™ FOTO/UV 15, 21 and 26 Transilluminators
For excitation of DNA gels containing UV-sensitive stains/dyes such as ethidium bromide (EtBR) and EtBR-substitutes
UVP UV/White Light 8-Watt Transilluminators
Newly-designed for better performance and efficiency
UviClear™ UV Transparent Glass Protectors
Replacement part for Fotodyne UV transilluminators
Spectroline™ Select™ Series UV Transilluminators
Economically priced transilluminators with high-clarity fluorescent response, sensitivity to 1 to 2ng, 312nm wavelength, variable filter areas and Outstanding irradiance uniformity.
UVP Visi-Blue™ Transilluminators
UV-safe units convert 365nm UV to 460-470nm blue light for use with various stains including GFP stains as well as SYBR Green, and SYPRO Orange stains. The amber protective cover blocks blue light transmission and allows visualization of most stains above 500nm.
Slimline™ UV Transilluminator Products for Science Education
For photodocumentation of small electrophoresis gels.
Spectroline™ Bi-O-Vision™ Series White Light Transilluminators
Offers both medium wave UV (312nm) and white light for dual-light convenience in one box
UV Blocking Covers
Replacement parts for Fotodyne UV Transilluminators
Fotodyne™ FOTO/Phoresis™ UV Transilluminator
Patented UV Transilluminator for Mini Gels; ideal for safe visualization and photography of DNA gels without leaving the lab bench.
UVP Model M-10E Mini UV Transilluminators: Filter Assembly
Accessory for UVP Model M-10E Mini UV Transilluminators
UVP White Light Transilluminator
Supplying back-lit white light
Spectroline™ Ultraviolet Transilluminators
Typical peak UV intensity of 9000 to 6000μw/cm2 (6000 to 4500μw/cm2 on dual-light models) produces brilliant fluorescence and high clarity
  spinner