Balances and Weighing
3B Scientific™ Brass Weights Set Products for Science Education
Set of nine hooked brass weights in storage case.
EISCO Compression Balance Products for Science Education
Durable plastic body.
EISCO Dual Scale Spring Scales Products for Science Education
Metric and Newton ranges.
EISCO Demo Lever Balance Products for Science Education
For demonstration of static forces, levers and an analytical balance.
Fractional Brass Masses Products for Science Education
Mass set meets weighing standards.
Stainless-Steel Weight Set Products for Science Education
Each polished, stainless-steel weight is inscribed with its capacity.
Black Enamel Hooked Weight Set Products for Science Education
Cast iron weights with black enamel finish can be hooked in tandem.
OHAUS™ Spring Scales: Pull Type Products for Science Education
Designed to stand up to heavy student use and withstand sudden load releases without breaking. Mettler Toledo Pull Type Spring Scales are made from rugged shatterproof plastic and metal. They are color-coded for easy identification of capacity.
OHAUS™ Harvard Trip Balances Products for Science Education
Ideal for comparative weighing programs and centrifuge tube balancing. OHAUS Harvard Trip Balances employ a two-pan design based on the classic Roberval balance principle. They are commonly used to determine the difference in mass between two objects, rather than their absolute value.
OHAUS™ Spring Scales: Dial Type Products for Science Education
Designed to stand up to heavy student use and withstand sudden load releases without breaking. OHAUS Dial Type Spring Scales have rugged shatterproof plastic and metal construction.
OHAUS™ Triple Beam Pro Balance Products for Science Education
Accurate, dependable and stylish with superior quality and durability. OHAUS Triple Beam Pro Balance is an ideal learning tool to help visualize mass measurement principles and to emphasize a hands-on learning approach.
OHAUS™ Vinyl Dust Covers for Triple Beam Balances Products for Science Education
For use with triple beam balances.
OHAUS™ Triple Beam Balances Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Accurate, rugged and dependable. OHAUS Triple Beam Balances have stainless-steel weighing pans and bearing covers which prevent rust and corrosion.
OHAUS™ Compact Scales Products for Science Education
Rugged construction, easy operation and great performance make these scales a truly remarkable value.
OHAUS™ Primer Balance Products for Science Education
Colorful, easy-to-clean, and very economical!
OHAUS™ Hand-Held Scales Products for Science Education
Convenient, compact and portable weighing capability in the palm of your hand. OHAUS Hand-Held Scales are small scales with sleek styling and convenient size.
Specific Gravity Rod and Clamp Assembly Products for Science Education
For use with Triple Beam Mechanical balances.
OHAUS™ Gold Series Pocket Scales Products for Science Education
Perfect for weighing jewelry, coins and small parts. OHAUS Gold Series Pocket Scales have large backlit LCD display and external calibration.
OHAUS™ Cent-O-Gram™ and Dial-O-Gram™ Balances Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Support centigram accuracy and encourage student precision. OHAUS Cent-O-Gram and Dial-O-Gram Balance comes with three-point base, special floating bearing principle, and beam design which eliminates need for level adjustment.
OHAUS™ Triple Pro Mechanical Balance Products for Science Education
Balance has integral covered mass storage, including weight set and built in handle for ease of use.
OHAUS™ RS232 and USB Interface Kits Products for Science Education
For use with Ohaus Explorer and Navigator XL balances.
OHAUS™ 1050mL Bowl For CS Series Compact Scales Products for Science Education
For use with CS Series Compact Scales.
Security Device for Pioneer and Explorer Series Balances Products for Science Education
For use with Pioneer and Explorer series balances.
OHAUS™ Portable Culinary Scale: Model CL5000F Products for Science Education
Combines precision measurement, easy-to-clean design and portability in one affordable package.
OHAUS™ Explorer™ Series Analytical Balances Products for Science Education
Faster weighing with improved repeatability and vibration filtering.
OHAUS™ Pocket Scales Products for Science Education
Great performance in a unique yet functional design.
OHAUS™ Valor™ 3000 Xtreme Portable Scales Products for Science Education
Compact portioning scales offer high precision and extreme durability for the harshest environments.
OHAUS™ Security Lock-down Device for Explorer™ Pro Toploading and Analytical Balances Products for Science Education
For use with Explorer Pro Toploading and Analytical Balances.
  spinner