Balance Software
Mettler Toledo™ Quantos Dosing Heads Available on GSA/VA Contract for Federal Government customers only.
Intelligent dosing heads for precise dosing of free flowing powdery substances such as APIs or reference standards where purity and quality of substance is essential. Mettler Toledo Quantos Dosing Head includes an integrated stirrer which guarantees a homogeneous flow of the powder.
Mettler Toledo™ Terminal Extension Cable for Analytical Balances Available on GSA/VA Contract for Federal Government customers only.
For use with XP, XS and XA analytical balances.
Mettler Toledo™ Balance Printer Accessories: Cables
Printer accessories for use with Mettler Toledo balances.
Mettler Toledo™ Temperature Calibration Kits Available on GSA/VA Contract for Federal Government customers only.
For use with halogen moisture analyzers. Mettler Toledo Temperature Calibration Kits are used to adjust halogen drying units to ensure consistent results.
Mettler Toledo™ LabX™ Balance Software Available on GSA/VA Contract for Federal Government customers only.
Optional licenses that can be added to LabX installation.
Mettler Toledo™ ErgoClips™ Small Basket For Weighing Available on GSA/VA Contract for Federal Government customers only.
For use with test tubes, centrifuge tubes or small plastic tubes of XP, XS, XA analytical and XP, XS micro balance.
Mettler Toledo™ Protective Covers for for NewClassic ME Balances
Protective covers for NewClassic ME balances.
Mettler Toledo™ Antistatic Systems Available on GSA/VA Contract for Federal Government customers only.
U-shaped and Point Ionizer antistatic systems for discharging electrostatically charged samples prior to weighing. Mettler Toledo Antistatic Systems use a high voltage to generate positively and negatively charged ions which are attracted by the electrostatically charged object and neutralize the disruptive charge on the surface of the weighing sample. As a result, the forces that distort the weighing result also disappear.
Mettler Toledo™ LabX™ Balance Express Software Available on GSA/VA Contract for Federal Government customers only.
Weighs different classes of material with improved documentation allowing electronic integration of weights and sample IDs with other software systems within lab.
Mettler Toledo™ LocalCan Interface for XS Series Analytical Balances Available on GSA/VA Contract for Federal Government customers only.
LocalCan Interface for XS Series Analytical Balances.
Mettler Toledo™ LabX™ Direct Balance Software Available on GSA/VA Contract for Federal Government customers only.
Increases the efficiency and accuracy of weighing applications. Fisher Scientific - Mettler Toledo LabX Direct Balance Software is an easy-to-use software designed to transfer data from one balance into an open application like an Excel™ spreadsheet.
Mettler Toledo™ SmartCal™ Moisture Balance Reference Substances Available on GSA/VA Contract for Federal Government customers only.
Verify the performance of your moisture analyzer in just 10 minutes. Mettler Toledo SmartCal Moisture Balance Reference Substances can be run like any other sample to fulfill demands of industry regulation and company audits at the same time.
Mettler Toledo™ ErgoClips™ Weighing Kit Available on GSA/VA Contract for Federal Government customers only.
For use with XP, XS, XA analytical and XP, XS micro balance.
Mettler Toledo™ ErgoClips™ Titration Basket
For use with 100mL Titration beakers, test tubes of XP, XS, XA analytical and XP, XS micro balance.
Mettler Toledo™ ErgoClips™ Stainless-steel Stand Available on GSA/VA Contract for Federal Government customers only.
For use with XS and XP analytical balance.
Mettler Toledo™ Weighing Pan for NewClassic ME Balances Available on GSA/VA Contract for Federal Government customers only.
Accessory for use with NewClassic ME Balances.
Mettler Toledo™ Dust Covers for NewClassic ME Balance
For use with NewClassic ME Balances.
Mettler Toledo™ ErgoClips™ Weighing Boat Holder Available on GSA/VA Contract for Federal Government customers only.
For use with weighing boat of XP, XS, XA analytical and XP, XS micro balance.
Mettler Toledo™ ErgoClips™ Small Flask Available on GSA/VA Contract for Federal Government customers only.
For use with XP, XS, XA analytical and XP, XS micro balance.
Mettler Toledo™ Transport Case for MS-S Balances Available on GSA/VA Contract for Federal Government customers only.
Accessory for use with MS-S Balances.
Mettler Toledo™ ErgoClips™ Solution Kits Available on GSA/VA Contract for Federal Government customers only.
Allow easy weighing directly into tare containers. Mettler Toledo ErgoClips Solution Kit includes all necessary tools for direct dosing with storage case.
Mettler Toledo™ ErgoClips™ Flask For Weighing Available on GSA/VA Contract for Federal Government customers only.
For use with XP, XS, XA analytical and XP, XS micro balance.
Mettler Toledo™ Virtual Infrared COM-Port Driver for SevenGo™ Pro Meters
Creates a virtual infrared COM-port for IR-USB adapter.
Mettler Toledo™ Damping Element
For underneath marble pad
  spinner