Immersion Circulators
Thermo Scientific™ AC150 Immersion Circulators Available on GSA/VA Contract for Federal Government customers only.
AC150 is ideal when applications require greater pumping performance, ramp programming, application alarms and temperature ranges from −50° to +150°C.
ARCTIC Refrigerated/Heated Bath SC100 A10 - 230V
Sophisticated digital control technology designed to ensure accuracy and reproducibility of your liquid temperature control procedures
Thermo Scientific™ Richard-Allan Scientific™ Resolve™ Immersion Oil Available on GSA/VA Contract for Federal Government customers only.
Eliminate toxicity and disposal problems with the Thermo Scientific™ Richard-Allan Scientific™ Resolve™ Immersion Oil.
Thermo Scientific™ PC201 Immersion Circulators Available on GSA/VA Contract for Federal Government customers only.
These circulators feature a 3.0Kw heater for rapid heatup, an all stainless-steel pump with ceramic rotors and extreme temperature performance ranging from −50° to +200°C.
Thermo Scientific™ AC200 Immersion Circulator
Ideal when applications require greater pumping performance, ramp programming, application alarms, and temperature ranges from −50° to +200°C
Thermo Scientific™ SC150 Immersion Circulators Available on GSA/VA Contract for Federal Government customers only.
Maintain temperature from −28° to +150°C, with SC150 Immersion Circulators—a basic model in a new generation of superior quality thermostats.
Thermo Scientific™ SC150L Immersion Circulators Available on GSA/VA Contract for Federal Government customers only.
Maintain temperature from −28° to +150°C with this superior thermostat featuring an increased immersion depth to accomodate larger or deeper baths.
Thermo Scientific™ SC100 Immersion Circulators Available on GSA/VA Contract for Federal Government customers only.
SC100 is one of the basic models in the new generation of superior quality thermostats that maintain temperature from −28° to +100°C.
Thermo Scientific™ AC200 Immersion Circulators Available on GSA/VA Contract for Federal Government customers only.
AC200 is ideal when applications require greater pumping performance, ramp programming, application alarms, and temperature ranges from −50° to +200°C.
Thermo Scientific™ PC200 Immersion Circulators Available on GSA/VA Contract for Federal Government customers only.
These circulators are ideal for applications that require sophisticated control, multiple ramp programming and extreme temperature performance ranging from −50° to +200°C.
Fisher Scientific™ Isotemp™ Heated Immersion Circulators: Model 6200B Available on GSA/VA Contract for Federal Government customers only.
Convenient, simple way to convert open baths to constant temperature baths.
Fisher Scientific™ Isotemp™ Heated Immersion Circulators: Model 5150B Available on GSA/VA Contract for Federal Government customers only.
Convenient, simple way to convert open baths to constant temperature baths.
Fisher Scientific™ Isotemp™ Heated Immersion Circulators: Model 5150C Available on GSA/VA Contract for Federal Government customers only.
Convenient, simple way to convert open baths to constant temperature baths.
Fisher Scientific™ Isotemp™ Heated Immersion Circulators: Model 4100C Available on GSA/VA Contract for Federal Government customers only.
Convenient, simple way to convert open baths to constant temperature baths.
Fisher Scientific™ Isotemp™ Heated Immersion Circulators: Model 4100B Available on GSA/VA Contract for Federal Government customers only.
Convenient, simple way to convert open baths to constant temperature baths.
Carl Zeiss Immersol™ Immersion Oil
For Carl Zeiss Microscopes
Cargille Immersion Oil Available on GSA/VA Contract for Federal Government customers only.
Greatly increases light-gathering power with high-power objectives
Cargille Immersion Oil Products for Science Education
Finest immersion oil available.
  spinner